Skip to Content
Merck
All Photos(4)

Key Documents

C55402

Sigma-Aldrich

4-Chloro-3-methylphenol

99%

Synonym(s):

4-Chloro-m-cresol, PCMC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CH3)OH
CAS Number:
Molecular Weight:
142.58
Beilstein:
1237629
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

solid

bp

235 °C (lit.)

mp

63-65 °C (lit.)

SMILES string

Cc1cc(O)ccc1Cl

InChI

1S/C7H7ClO/c1-5-4-6(9)2-3-7(5)8/h2-4,9H,1H3

InChI key

CFKMVGJGLGKFKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chloro-3-methylphenol, also known as p-chlorocresol or chloroxylenol, is commonly used as a building block in the synthesis of organic compounds. [1] It can be utilized to synthesize various biologically active compounds, including pharmaceuticals and agrochemicals.[2] (2) Additionally, it serves as a potent disinfectant and antiseptic and is also used as a preservative in medicinal products.[3]

Application

  • Compression of mono-4 chloro, 3-methylphenyl phosphate ester and di-4 chloro, 3-methylphenyl phosphate ester through kinetic study: This study discusses a method using 4-chloro-3-methylphenol and POCl3 taken in a 2:1 ratio, exploring their kinetic behaviors (AR Teli, AA Khan, SH Dar, A Rashid, 2017).
  • Commercially Important Chlorinated Phenols: Reviews the usage of 4-chloro-3-methylphenol among other chlorinated phenols in various industrial applications (K Smith, G El-Hiti).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

244.4 °F - closed cup

Flash Point(C)

118.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic and natural phenols
Tyman et al.
Elsevier's Integrated Review Biochemistry (Second Edition) (1996)
Synthesis and insecticidal activity evaluation of virtually screened phenylsulfonamides
Gang et al.
Journal of Agricultural and Food Chemistry, 68, 11665-11671 (2020)
Synthesis and biological evaluation of novel aryloxyacetic acid hydrazide derivatives as anticancer agents
Sevil et al.
Synthetic Communications, 51, 2634-2643 (2021)
Ramón Jiménez-Moreno et al.
Pflugers Archiv : European journal of physiology, 459(5), 725-735 (2010-01-14)
The endoplasmic/sarcoplasmic reticulum (ER/SR) plays a crucial role in cytoplasmic signalling in a variety of cells. It is particularly relevant to skeletal muscle fibres, where this organelle constitutes the main Ca2+ store for essential functions, such as contraction. In this
Andrew P Ziman et al.
Biophysical journal, 99(8), 2705-2714 (2010-10-21)
Skeletal muscle stores Ca²(+) in the sarcoplasmic reticulum (SR) and releases it to initiate contraction, but the concentration of luminal Ca²(+) in the SR ([Ca²(+)](SR)) and the amount that is released by physiological or pharmacological stimulation has been difficult to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service