Skip to Content
Merck
All Photos(1)

Key Documents

32053

Supelco

Bronopol

PESTANAL®, analytical standard

Synonym(s):

2-Bromo-2-nitro-1,3-propanediol, BNPD, BNPK, Bronopol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2C(Br)(NO2)CH2OH
CAS Number:
Molecular Weight:
199.99
Beilstein:
1705868
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

130-133 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

OCC(Br)(CO)[N+]([O-])=O

InChI

1S/C3H6BrNO4/c4-3(1-6,2-7)5(8)9/h6-7H,1-2H2

InChI key

LVDKZNITIUWNER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bronopol is a broad-spectrum antibacterial agent. It is mostly used as a preservative in cosmetic, toiletry etc. It is also used in aerosol formulations.1

Application

Bronopol has been used as reference standard in ultra performance liquid chromatography (UPLC) coupled to inductively coupled plasma mass spectrometry (UPLC-ICP-MS) method for determination of bromine containing preservatives from cosmetic products.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyphenation of ultra performance liquid chromatography (UPLC) with inductively coupled plasma mass spectrometry (ICP-MS) for fast analysis of bromine containing preservatives.
Bendahl L et.al.
Journal of Pharmaceutical and Biomedical Analysis, 40(3), 648-652 (2006)
Sara M Picón-Camacho et al.
Veterinary parasitology, 186(3-4), 237-244 (2011-12-14)
Ichthyophthirius multifiliis Fouquet, 1876 infections on intensively reared fish stocks can increase rapidly, which if left unmanaged, can result in the heavy loss of stock. The present study explores the efficacy of long duration, low dose (1, 2 and 5
D M Barbano et al.
Journal of dairy science, 93(12), 6000-6011 (2010-11-26)
Our objective was to determine the effect of commonly used milk preservatives on the accuracy of fat, protein, and lactose content determination in milk by mid-infrared (mid-IR) milk analysis. Two producer raw milks (Holstein and Jersey) and 2 pasteurized modified
Mustapha Kireche et al.
Contact dermatitis, 63(4), 192-202 (2010-08-25)
Compounds slowly releasing formaldehyde, the so-called formaldehyde-releasers, are commonly employed as preservatives in cosmetics instead of free formaldehyde, which is a strong skin sensitizer. It has been long accepted that formaldehyde-releaser sensitization is attributable to released formaldehyde. However, clinical studies
Mustapha Kireche et al.
Chemical research in toxicology, 24(12), 2115-2128 (2011-11-01)
Formaldehyde and formaldehyde releasers are widely used preservatives and represent an important group of skin sensitizers. Formaldehyde is very often suspected to be the sensitizing agent of formaldehyde-releasers; however, many reported clinical cases of contact allergy to these molecules such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service