About This Item
Recommended Products
grade
analytical standard
vapor density
4.9 (vs air)
vapor pressure
1 mmHg ( 16.5 °C)
3.77 mmHg ( 37.7 °C)
CofA
current certificate can be downloaded
autoignition temp.
410 °F
expl. lim.
2.6 %
packaging
ampule of 1000 mg
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
refractive index
n20/D 1.411 (lit.)
bp
174 °C (lit.)
mp
−30 °C (lit.)
density
0.73 g/mL at 25 °C (lit.)
application(s)
environmental
petroleum
format
neat
storage temp.
2-30°C
SMILES string
CCCCCCCCCC
InChI
1S/C10H22/c1-3-5-7-9-10-8-6-4-2/h3-10H2,1-2H3
InChI key
DIOQZVSQGTUSAI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
It may be used as an extraction solvent of headspace single-drop microextraction (HS-SDME) with droplet derivatization for the determination of hexanal and heptanal in human blood by GC-MS.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Asp. Tox. 1 - Flam. Liq. 3
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
114.8 °F - closed cup
Flash Point(C)
46.0 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service