Z3902
Zerumbone
≥98% (HPLC)
Synonym(s):
(2E,6E,10E)-2,6,9,9-tetramethylcycloundeca-2,6,10-trien-1-one, Zerumbone (6CI,7CI)
About This Item
Recommended Products
Assay
≥98% (HPLC)
form
powder
color
white to off-white
solubility
DMSO: ≥10 mg/mL
storage temp.
2-8°C
SMILES string
C\C1=C/CC(C)(C)\C=C\C(=O)\C(C)=C\CC1
InChI
1S/C15H22O/c1-12-6-5-7-13(2)14(16)9-11-15(3,4)10-8-12/h7-9,11H,5-6,10H2,1-4H3/b11-9+,12-8+,13-7+
InChI key
GIHNTRQPEMKFKO-SKTNYSRSSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- for liquid chromatography with mass spectrometry (LC-MS-MS)
- to investigate its potential application in the prevention
- treatment of esophageal squamous cell carcinomas (ESCC)
Biochem/physiol Actions
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service