Skip to Content
Merck
All Photos(2)

Key Documents

S6317

Sigma-Aldrich

L-Sulforaphane

≥95% (HPLC), oil

Synonym(s):

(R)-1-Isothiocyanato-4-(methylsulfinyl)butane, 4-Methylsulfinylbutyl isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NOS2
CAS Number:
Molecular Weight:
177.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95% (HPLC)

form

oil

optical activity

[α]/D -70 to -90°, c = 1.0 in chloroform-d

color

light yellow

solubility

DMSO: >5 mg/mL

storage temp.

−20°C

SMILES string

CS(=O)CCCCN=C=S

InChI

1S/C6H11NOS2/c1-10(8)5-3-2-4-7-6-9/h2-5H2,1H3/t10-/m1/s1

InChI key

SUVMJBTUFCVSAD-SNVBAGLBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Sulforaphane was used to study Nrf2-mediated signaling in mouse primary preadipocytes6 and murine macrophage RAW 264.7 cell line.7

Biochem/physiol Actions

L-Sulforaphane is a potent, selective inducer of phase II detoxification enzymes with anticarcinogenic properties. Organosulfur compound found in cruciferous vegetables, including broccoli.
Sulforaphane is an anti-cancer, anti-microbial and anti-diabetic compound found in cruciferous vegetables.3,4 It induces the production of detoxifying enzymes such as quinone reductase and glutathione S-transferase that cause xenobiotic transformation. Sulforaphane also increases the transcription of tumor suppressor proteins and inhibits histone deacetylases. It modulates inflammatory responses by suppressing the LPS-mediated expression of iNOS, COX-2, IL-1β and TNF-α.3,5

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tiziana Latronico et al.
Inflammopharmacology, 29(2), 561-571 (2020-11-17)
Isothiocyanates (ITCs), present as glucosinolate precursors in cruciferous vegetables, have shown anti-inflammatory, antioxidant and anticarcinogenic activities. Here, we compared the effects of three different ITCs on ROS production and on the expression of matrix metalloproteinase (MMP)-2 and -9, which represent
Pengfei Liu et al.
Redox biology, 38, 101766-101766 (2020-10-31)
Idiopathic pulmonary fibrosis (IPF) is a progressive and irreversible disease characterized by an increase in differentiation of fibroblasts to myofibroblasts and excessive accumulation of extracellular matrix in lung tissue. Pharmacological activation of NRF2 has proved to be a valuable antifibrotic
Mohammed Ali Selo et al.
Frontiers in bioengineering and biotechnology, 8, 1030-1030 (2020-10-06)
Multidrug resistance-associated protein-1 (MRP1/ABCC1) is highly expressed in human lung tissues. Recent studies suggest that it significantly affects the pulmonary disposition of its substrates, both after pulmonary and systemic administration. To better understand the molecular mechanisms involved, we studied the
Valeria Capurro et al.
PloS one, 8(2), e56870-e56870 (2013-02-27)
Alzheimer's disease (AD) is characterized by progressive loss of cognitive function, dementia and altered behavior. Over 30 million people worldwide suffer from AD and available therapies are still palliative rather than curative. Recently, Memoquin (MQ), a quinone-bearing polyamine compound, has
Morgane De Tollenaere et al.
Journal of cosmetic dermatology, 19(2), 444-455 (2019-06-25)
Several studies evidenced significant increase of cortisol is the consequence of UV or emotional stress and leads to various deleterious effects in the skin. The well-aging, a new concept of lifestyle, procures an alternative to the anti-aging strategy. We demonstrated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service