Y1896
Yeast Synthetic Drop-out Medium Supplements
without lysine
Synonym(s):
Yeast Synthetic Drop-out Media Supplements
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Looking for similar products? Visit Product Comparison Guide
Application
The selection of plasmids in yeast is based on the use of auxotrophic mutant strains that cannot grow without a specific media component (an amino acid, purine or pyrimidine). Transformation with a plasmid containing the mutated gene enables the transformant to grow on a medium lacking the required component. Sigma′s Yeast Synthetic Drop-Out Media Supplements create a richer medium for better yield and growth rate, and increase the probability of successful transformations when screening libraries or performing gene knock-outs.
Other Notes
Mixtures of amino acids and other nutrients to be added to Yeast Nitrogen Base Without Amino Acids.
related product
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service