Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Yeast Synthetic Drop-out Medium Supplements
Products
Product Category
media (10)
Brand
Sigma-Aldrich (8)
Physical Form
Quality Segment
Application
Technique
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
Y1501
Yeast Synthetic Drop-out Medium Supplements,
without uracil
Expand
View Pricing
Y1376
Yeast Synthetic Drop-out Medium Supplements,
without leucine
Expand
View Pricing
Y2001
Yeast Synthetic Drop-out Medium Supplements,
without histidine, leucine, tryptophan and uracil
Expand
View Pricing
Y1771
Yeast Synthetic Drop-out Medium Supplements,
without uracil, leucine, and tryptophan
Expand
View Pricing
Y1876
Yeast Synthetic Drop-out Medium Supplements,
without tryptophan
Expand
View Pricing
Y1751
Yeast Synthetic Drop-out Medium Supplements,
without histidine
Expand
View Pricing
Y0750
Yeast Synthetic Drop-out Medium Supplements,
without leucine and tryptophan
Expand
View Pricing
Y2146
Yeast Synthetic Drop-out Medium Supplements,
without histidine, leucine, and tryptophan
Expand
View Pricing
Y2021
Yeast Synthetic Drop-out Medium Supplements,
without histidine, leucine, tryptophan, and adenine
Expand
View Pricing
Y1896
Yeast Synthetic Drop-out Medium Supplements,
without lysine
Expand
View Pricing