Skip to Content
Merck
All Photos(1)

Key Documents

S1141

Sigma-Aldrich

Spermine tetrahydrochloride

BioReagent, for molecular biology

Synonym(s):

N,N′-Bis(3-aminopropyl)-1,4-butanediamine tetrahydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H26N4 · 4HCl
CAS Number:
Molecular Weight:
348.18
Beilstein:
3911771
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.52

grade

Molecular Biology
for molecular biology

product line

BioReagent

form

solid

mp

310-311 °C (dec.) (lit.)

solubility

H2O: soluble 100 mg/mL, clear, colorless to light yellow

foreign activity

DNase, RNase, protease, NICKase, none detected

storage temp.

room temp

SMILES string

Cl.Cl.Cl.Cl.NCCCNCCCCNCCCN

InChI

1S/C10H26N4.4ClH/c11-5-3-9-13-7-1-2-8-14-10-4-6-12;;;;/h13-14H,1-12H2;4*1H

InChI key

XLDKUDAXZWHPFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Spermine is a polyamine involved in cellular metabolism and DNA stabilization. In molecular biology it can be used in buffers for purification of DNA.

Application

Used to precipitate DNA from low salt aqueous buffers.

Biochem/physiol Actions

Mixed NMDA agonist/antagonist at the polyamine site. Neuroprotective effects have been observed at high concentrations (1 mM), while neurotoxicity is observed at lower concentrations. It enhances agonist effectiveness at the strychnine-insensitive glycine site. Plays a role in cellular proliferation and differentiation; inhibits neuronal nitric oxide synthase (nNOS).
Mixed NMDA agonist/antagonist at the polyamine site. Plays a role in cellular proliferation and differentiation; inhibits neuronal nitric oxide synthase (nNOS).
Mixed NMDA agonist/antagonist at the polyamine site. Role in cellular proliferation and differentiation; inhibits neuronal nitric oxide synthase (nNOS).

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L L Vindeløv et al.
Cancer research, 40(11), 4295-4300 (1980-11-01)
Flow-cytometric DNA analysis yields information on ploidy and proliferative characteristics of a cell population. The analysis was implemented on small-cell anaplastic carcinoma of the lung using a rapid detergent technique for the preparation of fine-needle aspirates for DNA determination and
Binh V Vu et al.
PloS one, 7(12), e51863-e51863 (2013-01-04)
While most nucleic acids are intracellular, trace amounts of deoxyribonucleic acid (DNA) and ribonucleic acid (RNA), including micro RNAs, can also be found in peripheral blood. Many studies have suggested the potential utility of these circulating nucleic acids in prenatal
D M Hougaard et al.
The Biochemical journal, 238(1), 43-47 (1986-08-15)
Two recently developed fluorescence cytochemical methods, specific for spermidine and spermine, were used to localize polyamines in the endocrine pancreas. The polyamines were restricted to the insulin-producing beta-cells and were mainly associated with the secretory granules. Chemical polyamine determinations carried
Aracely A Lutes et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(24), 9910-9915 (2011-05-06)
Speciation in animals commonly involves an extrinsic barrier to genetic exchange followed by the accumulation of sufficient genetic variation to impede subsequent productive interbreeding. All-female species of whiptail lizards, which originated by interspecific hybridization between sexual progenitors, are an exception
Davis, L., et al.
Basic Methods in Molecular Biology, 120-120 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service