Skip to Content
Merck
All Photos(3)

Key Documents

XNAPR

Sigma-Aldrich

REDExtract-N-Amp Plant PCR Kit

sufficient for 1000 extractions, sufficient for 1000 amplifications

Synonym(s):

Plant direct PCR kit, Plant direct PCR kit with loading dye

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106303
NACRES:
NA.55

usage

sufficient for 1000 amplifications
sufficient for 1000 extractions
sufficient for 1000 reactions

Quality Level

feature

dNTPs included
hotstart

technique(s)

PCR: suitable

color

red

application(s)

agriculture

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The REDExtract-N-Amp Plant PCR Kits enable the rapid detection of plant pathogens by direct PCR (polymerase chain reaction). These kits contain all the reagents necessary to rapidly extract genomic DNA from plant leaves and amplify targets of interest by direct PCR. The REDExtract-N-Amp PCR ReadyMix contains a dye that acts as a tracking dye and allows for convenient direct loading of PCR reactions onto agarose gels for analysis.

Application

REDExtract-N-Amp Plant PCR Kit has been used to extract genomic DNA from leaves and mycelial lawns. It has also been used for polymerase chain reaction.

Features and Benefits

  • Single-step extraction of plant genomic DNA for PCR in less than 15 minutes
  • No freezing, mechanical disruption, organic extraction, column purification or precipitation required
  • Specially formulated PCR ReadyMix for use with extract
  • Hot Start antibody for highly specific PCR amplification of genomic DNA
  • REDExtract-N-Amp requires no loading buffers or tracking dyes required for gel analysis
  • Compatible with high-throughput requirements for genetic analysis of plants
  • Extract stable at 4 °C for at least 6 months

Components

Kits contain Dilution solution, Extraction solution, and REDExtract-N-Amp PCR Reaction Mix.

Other Notes

For additional information, please see www.sigma-aldrich.com/extract-n-amp.

Legal Information

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser′s own internal research. No other patent rights (such as 5′ Nuclease Process patent rights) are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Antibody licensed for in vitro research use under U.S. Patent No. 5,338,671 and 5,587,287, and corresponding patents in other countries.

Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: 5,789,224, 5,618,711, 6,127,155 and claims outside the US corresponding to expired US Patent No. 5,079,352. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
REDExtract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martha E Trujillo et al.
The ISME journal, 4(10), 1265-1281 (2010-05-07)
Our current knowledge of plant-microbe interactions indicate that populations inhabiting a host plant are not restricted to a single microbial species but comprise several genera and species. No one knows if communities inside plants interact, and it has been speculated
José F Cobo-Díaz et al.
Systematic and applied microbiology, 37(3), 177-185 (2013-11-26)
Genista versicolor is an endemic legume from Sierra Nevada National Park which constitutes one of the UNESCO-recognized Biosphere Reserves. In the present study, a collection of strains nodulating this legume was analysed in characteristic soils of this ecosystem. Most strains
Sandra J Hey et al.
Plant biotechnology journal, 4(2), 219-229 (2006-12-21)
The regulation of phytosterol biosynthesis in seeds is of interest to biotechnologists because of the efficacy of dietary phytosterols in reducing blood cholesterol in humans. Mevalonate synthesis via 3-hydroxy-3-methylglutaryl-coenzyme A reductase (HMG-CoA reductase) is a key step in phytosterol biosynthesis.
Cynthia E Gallagher et al.
Plant physiology, 135(3), 1776-1783 (2004-07-13)
Despite ongoing research on carotenoid biosynthesis in model organisms, there is a paucity of information on pathway regulation operating in the grasses (Poaceae), which include plants of world-wide agronomic importance. As a result, efforts to either breed for or metabolically
Rowan S McKibbin et al.
Plant biotechnology journal, 4(4), 409-418 (2006-12-21)
Transgenic potato (Solanum tuberosum cv. Prairie) lines were produced over-expressing a sucrose non-fermenting-1-related protein kinase-1 gene (SnRK1) under the control of a patatin (tuber-specific) promoter. SnRK1 activity in the tubers of three independent transgenic lines was increased by 55%-167% compared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service