Skip to Content
Merck
All Photos(6)

Key Documents

WH0003146M8

Sigma-Aldrich

Monoclonal Anti-HMGB1 antibody produced in mouse

clone 2F6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DKFZp686A04236, Anti-HMG1, Anti-HMG3, Anti-SBP1, Anti-high-mobility group box 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2F6, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HMGB1(3146)

General description

High mobility group box 1 (HMGB1) is a DNA binding protein, consisting of negatively charged amino acids in C-terminus and two DNA binding motifs, called BOX A and B. HMGB1 is mapped to human chromosome 13q12.3. Necrotic cells and neuritis express HMGB1. Monocytes and macrophages upon activation release HMGB1.

Immunogen

HMGB1 (NP_002119, 1 a.a. ~ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MGKGDPKKPRGKMSSYAFFVQTCREEHKKKHPDASVNFSEFSKKCSERWKTMSAKEKGKFEDMAKADKARYEREMKTYIPPKGETKKKFK

Application

Monoclonal Anti-HMGB1 antibody produced in mouse has been used in the detection of HMGB1 in pancreatic ductal adenocarcinogenic (PDAC) cell lines using fluorescent microscopy. and in western blotting.

Biochem/physiol Actions

High mobility group box 1 (HMGB1) functions to stabilize nucleosome and also regulates gene transcription. The phosphorylation of the nuclear localization signal in HMGB1 regulates its transport between cytoplasm and nucleus. It assists in nucleosome remodeling by eliciting chaperone functionality. Inflammation triggers high expression of HMGB1 and its inhibition may be useful for treating refractory M. pneumoniae pneumonia (RMPP). Elevated levels of HMGB1 is present in cardiovascular diseases. HMGB1 regulates autophagy in human liver cells in absence of oxygen followed reperfusion. High levels HMGB1 expression is seen in adenocarcinoma (AC), gall bladder and squamous cell/adenosquamous (SC/ASC) cancer. Polymorphisms in HMGB1 is present in oral squamous cell carcinoma (OSCC).

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High expression of HMGB1 in children with refractory Mycoplasma pneumoniae pneumonia
Ding Y, et al.
BMC Infectious Diseases, 18(1), 439-439 (2018)
A functional variant at the miRNA binding site in HMGB1 gene is associated with risk of oral squamous cell carcinoma
Lin CY, et al.
Oncotarget, 8(21), 34630-34630 (2017)
Correlation of HMGB1 expression to progression and poor prognosis of adenocarcinoma and squamous cell/adenosquamous carcinoma of gallbladder
Shi Z, et al.
American Journal of Translational Research, 7(10), 1532-1537 (2015)
Yam Nath Paudel et al.
Frontiers in pharmacology, 11, 613009-613009 (2021-03-19)
Epilepsy is a chronic brain disease afflicting around 70 million global population and is characterized by persisting predisposition to generate epileptic seizures. The precise understanding of the etiopathology of seizure generation is still elusive, however, brain inflammation is considered as
Perspectives on RAGE signaling and its role in cardiovascular disease
Cohen Jr MM
American Journal of Medical Genetics. Part A, 161(11), 2750-2755 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service