Skip to Content
Merck
All Photos(5)

Key Documents

SAB1403925

Sigma-Aldrich

Monoclonal Anti-HMGB1 antibody produced in mouse

clone 1D5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp686A04236, HMG1, HMG3, SBP-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1D5, monoclonal

form

buffered aqueous solution

mol wt

antigen ~49.76 kDa

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HMGB1(3146)

Immunogen

HMGB1 (AAH03378.1, 1 a.a. ~ 215 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MGKGDPKKPRGKMSSYAFFVQTCREEHKKKHPDASVNFSEFSKKCSERWKTMSAKEKGKFEDMAKADKARYEREMKTYIPPKGETKKKFKDPNAPKRPPSAFFLFCSEYRPKIKGEHPGLSIGDVAKKLGEMWNNTAADDKQPYEKKAAKLKEKYEKDIAAYRAKGKPDAAKKGVVKAEKSKKKKEEEEDEEDEEDEEEEEDEEDEDEEEDDDDE

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhangya Pu et al.
Journal of translational medicine, 20(1), 212-212 (2022-05-14)
Hepatocellular carcinoma (HCC) is the most common pathological type of liver cancer. Valosin-containing protein (VCP) is a member of the AAA-ATPase family associated with multiple molecular functions and involved in tumor metastasis and prognosis. However, the role of VCP in
David P J van Dijk et al.
Journal of cachexia, sarcopenia and muscle, 10(1), 123-130 (2018-11-01)
Most prognostic scoring systems for colorectal liver metastases (CRLMs) account for factors related to tumour biology. Little is known about the effects of the host phenotype to the tumour. Our objective was to delineate the relationship of systemic inflammation and
Si-Si Wang et al.
Journal of Cancer, 15(9), 2561-2572 (2024-04-05)
Purpose: Papillary thyroid cancer (PTC) stands as one of the most prevalent types of thyroid cancers, characterized by a propensity for in-situ recurrence and distant metastasis. The high mobility group protein (HMGB1), a conserved nuclear protein, plays a pivotal role
Maxine Bauzon et al.
Oncoimmunology, 8(4), e1565859-e1565859 (2019-03-25)
Oncology treatment has been revolutionized by the introduction of immune checkpoint inhibitor drugs, which enable 20-40% of patients to generate anti-tumor immune responses. Combination treatment approaches with chemotherapeutic drugs may enable responses in the remaining patient cohorts. In this regard
Keita Aoto et al.
Oncology reports, 39(1), 151-159 (2017-11-16)
It has been reported that chemo-radiotherapy can induce immunogenic tumor cell death (ICD), which triggers T-cell immunity mainly mediated by high-mobility group box 1 protein (HMGB1) and calreticulin. However, there is still limited information to support this theory relating to chemotherapy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service