W1500
Whey from bovine milk
spray-dried
Synonym(s):
Bovine Whey Protein
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.61
Recommended Products
biological source
bovine milk
Quality Level
form
spray-dried
concentration
≥11% protein (biuret)
technique(s)
ELISA: suitable
Gene Information
bovine ... MWHP(100531428)
General description
Whey proteins isolated from bovine milk plays a vital role in regulating immune responses. β-lactoglobulin is one of the highly expressed proteins in the bovine whey.
Application
Whey from bovine milk has been used:
- to test the efficacy of UV-C, nonthermal plasma and high intensity ultrasound treatments in reducing its allergenicity
- to determine its specific IgE, IgA and IgG subclass antibodies by sensitive enzyme-linked immunosorbent assay (ELISA) method
- as a component in the preparation of chitosan/whey protein (CWP) film forming solution
Physical form
Spray dried powder
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Chitosan/whey protein film as active coating to extend Ricotta cheese shelf-life
Di Pierro P, et al.
LWT--Food Science and Technology, 44(10), 2324-2327 (2011)
Determining the effect of UV-C, high intensity ultrasound and nonthermal atmospheric plasma treatments on reducing the allergenicity of alpha-casein and whey proteins
Tammineedi C, et al.
LWT--Food Science and Technology, 54(1), 35-41 (2013)
Effect of dietary whey protein concentrate on primary and secondary antibody responses in immunized BALB/c mice
Low PPL, et al.
International Immunopharmacology, 3(3), 393-401 (2003)
Tysheena P Charles et al.
PLoS pathogens, 17(2), e1009257-e1009257 (2021-02-09)
Stabilized HIV-1 envelope (Env) trimers elicit tier 2 autologous neutralizing antibody (nAb) responses in immunized animals. We previously demonstrated that BG505 SOSIP.664.T332N gp140 (BG505 SOSIP) immunization of rhesus macaques (RM) provided robust protection against autologous intra-vaginal simian-human immunodeficiency virus (SHIV)
Antonio Carroccio et al.
Scandinavian journal of gastroenterology, 41(4), 498-504 (2006-04-26)
Chronic constipation that is unresponsive to laxative treatment is a severe illness, but children unresponsive to laxatives have been successfully treated with an elimination diet. We report the first cases of refractory chronic constipation caused by food hypersensitivity in adults.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service