Skip to Content
Merck
All Photos(1)

Key Documents

M7409

Sigma-Aldrich

Nonfat-Dried Milk bovine

Synonym(s):

Nonfat milk powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

Quality Level

Application

Nonfat-Dried Milk bovine has been used as a blocking agent/solution in:
  • immunohistochemistry
  • western blotting
  • enzyme-linked immunosorbent assay (ELISA)

Reconstitution

Reconstitution with 100 mL deionized water yields a 3% w/v working solution.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Restituto Tocmo et al.
Free radical biology & medicine, 163, 102-115 (2020-12-15)
Xanthones from the tropical fruit mangosteen (Garcinia mangostana) display anti-inflammatory and anti-oxidative activities. Here, we isolate and identify potential inducers of the aryl hydrocarbon receptor (AhR) and nuclear factor erythroid 2-related factor 2 (Nrf2) signaling pathways from mangosteen using a
Mylène Boulay et al.
International journal of food microbiology, 335, 108903-108903 (2020-10-17)
Societal demand for plant-based foods is increasing. In this context, soya products fermented using lactic acid bacteria (LAB) are appealing because of their potential health and nutritional benefits. The thermophilic LAB Streptococcus thermophilus is an essential starter species in the
Xiaochuan Shan et al.
PeerJ, 6, e4661-e4661 (2018-04-24)
In 2015, as part of the Prostate Cancer Foundation-Movember Foundation Reproducibility Initiative, we published a Registered Report (Shan et al., 2015) that described how we intended to replicate selected experiments from the paper "Androgen Receptor Splice Variants Determine Taxane Sensitivity
Tayana Silva de Carvalho et al.
Molecular neurobiology, 56(12), 8477-8488 (2019-07-02)
Food composition influences stroke risk, but its effects on ischemic injury and neurological deficits are poorly examined. While severe reduction of protein content was found to aggravate neurological impairment and brain injury as a consequence of combined energy-protein malnutrition, moderate
Tayana Silva de Carvalho et al.
Translational stroke research, 11(1), 135-146 (2019-03-20)
Malnutrition predisposes to poor stroke outcome. In animal models, undernutrition protected against ischemic injury in some, but not in other studies. In view of diverse stroke models and food restriction paradigms, the consequences of undernutrition are poorly understood. Herein, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service