Skip to Content
Merck
All Photos(1)

Key Documents

V4765

Sigma-Aldrich

Valnoctamide

≥98% (NMR)

Synonym(s):

2-ethyl-3-methyl-pentanamide, Axiquel, Nirvanil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17NO
CAS Number:
Molecular Weight:
143.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (NMR)

form

powder

storage condition

desiccated

color

white to off-white

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

CCC(C)C(CC)C(N)=O

InChI

1S/C8H17NO/c1-4-6(3)7(5-2)8(9)10/h6-7H,4-5H2,1-3H3,(H2,9,10)

InChI key

QRCJOCOSPZMDJY-UHFFFAOYSA-N

Application

Valnoctamide has been used:
  • as a mood stabilizer to study its anti-cytomegalovirus (anti-CMV) effects in newborn mice brain
  • as a hypnotic sedative to study its cytotoxic effects on oligodendrocyte precursor cells (OPCs) and human oligodendroglioma cell line (HOG)
  • as a mood stabilizer to study its effects on inhibition of human cytomegalovirus

Biochem/physiol Actions

Valnoctamide exhibits anti-cytomegalovirus (anti-CMV) properties. It has therapeutic effects against status epilepticus (SE) and neuropathic pain. Valnoctamide also shows therapeutic effects against bipolar disorders.
Valproic acid (VPA) and derivatives such as valpromide and valnoctamide are anti-convulsant, mood stabilizing drugs, believed to function as indirect GABA agonists by inhibiting the transamination of GABA.

Features and Benefits

This compound is featured on the Glutamate/GABA Synthesis and Metabolism page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mood stabilizers inhibit cytomegalovirus infection
Ornaghi S, et al.
Virology, 499(29), 121-135 (2016)
M Bialer
Journal of controlled release : official journal of the Controlled Release Society, 62(1-2), 187-192 (1999-10-16)
Valproic acid (VPA) is one of the major antiepileptic drugs. However, its anticonvulsant potency is less than the other three major antiepileptic drugs. Furthermore, VPA causes two rare but severe side effects: teratogenicity and hepatotoxicity. We utilized pharmacokinetic considerations in
S Blotnik et al.
Drug metabolism and disposition: the biological fate of chemicals, 24(5), 560-564 (1996-05-01)
Valpromide (VPD) and valnoctamide (VCD) are amide derivatives of valproic acid (VPA), one of the major antiepileptic drugs (AEDs). In rodent models, both VPD and VCD are more potent as anticonvulsants than VPA. However, in humans, VPD served as a
Amidic derivatives of valproic acid as antiviral in HSV-1-infected oligodendrocytes.
Praena B, et al.
Antiviral Research, 54(29), 99-102 (2019)
F Pisani et al.
Epilepsia, 34(5), 954-959 (1993-09-01)
Six patients stabilized with carbamazepine (CBZ) therapy received an 8-day "add-on" supplement of valnoctamide (VCD), a tranquilizer available over the counter (OTC) in several European countries that exhibits promising anticonvulsant activity in animal models. During VCD intake, serum levels of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service