Skip to Content
Merck
All Photos(1)

Documents

T9822

Sigma-Aldrich

Anti-Tubulin, Polyglutamylated antibody, Mouse monoclonal

clone B3, purified from hybridoma cell culture

Synonym(s):

Anti-K-ALPHA-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

B3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~50 kDa

species reactivity

mouse, sea urchin (Lytechinus pictus and three others.), bovine, monkey, rabbit, human, rat, Oxyrrhis marina (dinoflagellate), chicken

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
microarray: suitable
western blot: 0.5-1 μg/mL using cytosolic fraction of rat brain extract

isotype

IgM

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Tubulin is a 100kD protein that is present in all cells as a heterodimer of two similar polypeptides α and β which assemble to form microtubules. Proper organization of microtubules is essential for several cellular functions like mitosis, meiosis, some forms of organellar movement, intracellular transport, flagellar movement and cytoskeletal functions. Monoclonal anti-tubulin, polyglutamylated antibody can be used as primary antibody in colocalization experiments. This antibody can also be used for studies involving the inhibition of flagellar motility in permeabilized sperm models. Mouse anti-tubulin, polyglutamylated antibody reacts specifically with the epitope present in the C-terminal region of α and β tubulins (glutamylated motif at amino acids 445-457 of α-tubulin).
Tubulin is a cellular colchicine-binding protein.

Immunogen

purified sea urchin (Lytechinus pictus) sperm axonemal proteins.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunocytochemistry (1 paper)
Western Blotting (1 paper)
Monoclonal Anti-Tubulin, Polyglutamylated antibody produced in mouse has been used in:
  • immunoprecipitation
  • immunohistochemistry
  • immunofluorescence

Monoclonal anti-tubulin, polyglutamylated antibody can be used in immunoblotting, immunocytochemistry and ELISA. It can also be used as primary antibodies (diluted 1:1000) in western blotting.

Biochem/physiol Actions

Microtubule plays an important role in DNA segregation.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitotic spindle assembly and chromosome segregation: refocusing on microtubule dynamics
Kline-Smith SL and Walczak CE
Molecular Cell, 15(3), 317-327 (2004)
A Mutation in CCDC50, a Gene Encoding an Effector of Epidermal Growth Factor-Mediated Cell Signaling, Causes Progressive Hearing Loss
Modamio-HoybjorS, et al.
American Journal of Human Genetics, 80(6), 1076-1089 (2007)
Kollu N Rao et al.
Biology open, 5(4), 424-428 (2016-03-05)
Mutations inRPGR(ORF15)(retinitis pigmentosa GTPase regulator) are a major cause of inherited retinal degenerative diseases. RPGR(ORF15)(1152 residues) is a ciliary protein involved in regulating the composition and function of photoreceptor cilia. The mutational hotspot in RPGR(ORF15)is an unusual C-terminal domain encoded
Karel Soucek et al.
The Prostate, 66(9), 954-965 (2006-03-17)
Multiple diverse posttranslational modifications of alpha-tubulin such as detyrosination, further cleavage of the penultimate glutamate residue (Delta2-tubulin), acetylation, and polyglutamylation increase the structural and functional diversity of microtubules. Herein, we characterized the molecular profile of alpha-tubulin posttranslational modifications in normal
Parathyroid hormone-related peptide represses chondrocyte hypertrophy through a protein phosphatase 2A/histone deacetylase 4/MEF2 pathway
Kozhemyakina E, et al.
Molecular and Cellular Biology, 29(21), 5751-5762 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service