Skip to Content
Merck
All Photos(4)

Key Documents

AB3201

Sigma-Aldrich

Anti-β-Tubulin III Antibody

CHEMICON®, rabbit polyclonal

Synonym(s):

Glu Tubulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Tubulin Antibody, Detyrosinated, Chemicon®, from rabbit

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, sea urchin, human, plant

should not react with

Saccharomyces cerevisiae

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TUBA1A(7846)

General description

Microtubules are involved in a wide variety of cellular activities ranging from mitosis and transport events to cell movement and the maintenance of cell shape. Tubulin itself is a globular protein which consists of two polypeptides (alpha and beta tubulin). Alpha and beta tubulin dimers are assembled to 13 protofilaments that form a microtubule of 22 nm diameter. Tyrosine ligase ads a C-terminal tyrosin to monomeric alpha tubulin. Assembled microtubules can again be detyrosinated by a cytoskeleton associated carboxypeptidase. Detyrosinated alpha tubulin is referred to as Glu-tubulin. Another post-translational modification of detyrosinated alpha tubulin is C-terminal polyglutamylation which is characteristic for microtubules in neuronal cells and the mitotic spindle.

Specificity

Expected to cross-react with mammals, sea urchin, plants, not yeast (Paturle-Lafanechere, L., 1991).
The antibody specifically recognizes the detyrosinated form (Glu tubulin) of the tubulin alpha chain (Bianchi, M., 2006; Goulimari, P., 2005).

Immunogen

Peptide corresponding to the C-terminal of Glu tubulin.

Application

Anti-Tubulin Antibody, Detyrosinated is an antibody against Tubulin for use in IC, IH(P) and WB.
Immunohistochemistry:
A previous lot was used on either frozen or paraffin embedded, fixed tissue (Gundersen, G.G., 1984): 1:200-1:500 dilution.

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
Cytoskeletal Signaling

Quality

Routinely evaluated by Western Blot on L6 lysate lysates.

Western Blot Analysis: 1:500 dilution of this lot detected detyrosinated tubulin on 10 μg of L6 lysate.

Target description

50 kDa

Physical form

Affinity purified immunoglobulin
Purified rabbit polyclonal in buffer containing liquid PBS.

Storage and Stability

Stable for 6 months at -20°C in undiluted aliquots from date of receipt.

Handling Recommendations:
Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Hela Whole Cell Lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunmei Li et al.
PLoS biology, 14(3), e1002416-e1002416 (2016-03-18)
Cilia have a unique diffusion barrier ("gate") within their proximal region, termed transition zone (TZ), that compartmentalises signalling proteins within the organelle. The TZ is known to harbour two functional modules/complexes (Meckel syndrome [MKS] and Nephronophthisis [NPHP]) defined by genetic
Carolyn Ott et al.
Cilia, 1(1), 3-3 (2013-01-29)
The central tenet of cilia function is sensing and transmitting information. The capacity to directly contact extracellular surfaces would empower primary cilia to probe the environment for information about the nature and location of nearby surfaces. It has been well
Joanna Lipka et al.
The EMBO journal, 35(3), 302-318 (2016-01-14)
In neurons, the polarized distribution of vesicles and other cellular materials is established through molecular motors that steer selective transport between axons and dendrites. It is currently unclear whether interactions between kinesin motors and microtubule-binding proteins can steer polarized transport.
Sunny Y Wong et al.
Nature medicine, 15(9), 1055-1061 (2009-08-25)
Primary cilia are present on most mammalian cells and are implicated in transducing Hedgehog (Hh) signals during development; however, the prevalence of cilia on human tumors remains unclear, and the role of cilia in cancer has not been examined. Here
Esther Castellano et al.
Nature communications, 7, 11245-11245 (2016-04-14)
RAS signalling through phosphoinositide 3-kinase (PI3-Kinase) has been shown to have an essential role in tumour initiation and maintenance. RAS also regulates cell motility and tumour invasiveness, but the role of direct RAS binding to PI3-Kinase in this remains uncertain.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service