Skip to Content
Merck
All Photos(2)

Key Documents

T3526

Sigma-Aldrich

Anti-TUBA1B (alpha-tubulin ubiquitous) Antibody

rabbit polyclonal

Synonym(s):

Anti-K-ALPHA-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-Tubulin antibody produced in rabbit, whole antiserum

biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

chicken, mouse

technique(s)

indirect immunofluorescence: 1:80 using cultured fibroblasts
western blot: 1:200 using chicken gizzard extract or mouse brain extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Microtubules are composed of tubulin and several additional proteins designated microtubule associated proteins (MAPs). Tubulin is a heterodimer which consists of an α-chain and a β-chain, both of which have a molecular weight of approximately 55,000. Microtubules form an elaborate cytoplasmic network during interphase and are reorganized into the mitotic spindle during cell division.
Tubulin is an extremely conserved and negatively charged protein. It is expressed in all eukaryotic cells, but mainly in brains.

Specificity

The product stains tubulin in cultured chicken fibroblasts or 3T3 mouse fibroblasts. Additional bands may be stained in immunoblotting.

Immunogen

microtubule proteins from brains of 13 day old chicken embryos

Application

Anti-Tubulin antibody produced in rabbit has been used in immunoblotting and indirect immunofluorescence.

Biochem/physiol Actions

αβ-tubulin heterodimers helps in formation of microtubules. Tubulin dimers exhibits GTPase activity, assembly and disassembly properties. It facilitates the construction of the flagellar apparatus.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Zhu et al.
Experimental and therapeutic medicine, 15(3), 2643-2648 (2018-02-20)
The aim of the present study was to investigate the mechanism of action by which naringin reverses the resistance of ovarian cancer cells to cisplatin. Semi-quantitative reverse transcription polymerase chain reaction (RT-PCR) and western blotting assays were used to detect
Microtubule proteins and their post-translational forms in the cerebrospinal fluid of patients with paraparesis associated with HTLV-I infection and in SH-SY5Y cells: an in vitro model of HTLV-I-induced disease
Maldonado H, et al.
Biological Research, 41(3), 239-259 (2008)
Tubulin and microtubule structure
Downing KH and Nogales E
Current Opinion in Cell Biology, 10(1), 16-22 (1998)
Isolating tubulin from nonneural sources
Methods in Cell Biology, 95, 17-32 (2010)
Tumor necrosis factor-related apoptosis-inducing ligand promotes migration of human bone marrow multipotent stromal cells
Secchiero P, et al.
Stem Cells, 26(11), 2955-2963 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service