Skip to Content
Merck
All Photos(1)

Key Documents

T9378

Sigma-Aldrich

Trypsin inhibitor

crude powder

Synonym(s):

Trypsin Inhibitor from lima beans

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Product Name

Trypsin inhibitor from Phaseolus limensis (lima bean), Type II-L, crude powder

biological source

Phaseolus limensis (lima bean)

type

Type II-L

form

crude powder

mol wt

9 kDa

solubility

0.067 M sodium phosphate buffer, pH 7.6: 1 mg/mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Trypsin inhibitor from Phaseolus limensis (lima bean) has been used:
  • in the inhibition of trypsin in human plasma
  • in the termination of proteolytic digestion in rabbit skeletal muscles
  • in the inhibition of salivary glands enzyme extract from Lygus lineolaris
  • in the inhibition of proteolytic activity in Agave tequilana enzyme extract

Trypsin inhibitor has been used as an affinity ligand for isolation of elastase-type enzymes.

Biochem/physiol Actions

Trypsin inhibitor from lima beans belongs to Bowman-Birk family of protease inhibitors. It is 9 kDa in molecular weight and contains nine disulfide bridges and is highly thermostable. It has trypsin and chymotrypsin binding subdomains.

Physical properties

Monomer has an apparent molecular weight of approx. 9,000 Da but undergoes a concentration and pH-dependent dimerization.

Unit Definition

One trypsin unit will produce a ΔA253 of 0.001 per min with BAEE as substrate at pH 7.6 at 25 °C; reaction volume 3.2 mL, 1 cm light path.

Preparation Note

Prepared by a modification of the method of Tauber, H., et al., J. Biol. Chem., 179, 1155 (1949) (modified).

Analysis Note

One mg will inhibit ≥0.8 mg of trypsin with activity of approx. 10,000 BAEE units per mg protein.

Other Notes

View more information on Trypsin Inhibitor.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In-house phase determination of the lima bean trypsin inhibitor: a low-resolution sulfur-SAD case
Debreczeni J, et al.
Acta crystallographica. Section D, Structural biology, 59(2), 393-395 (2003)
A Rapid and Reliable Method for Total Protein Extraction from Succulent Plants for Proteomic Analysis
Lledias F, et al.
The Protein Journal, 36(4), 308-321 (2017)
Partial characterization of trypsin-like protease and molecular cloning of a trypsin-like precursor cDNA in salivary glands of Lygus lineolaris
Zeng F, et al.
Comparative Biochemistry and Physiology. Part B, Biochemistry & Molecular Biology, 131(3), 453-463 (2002)
Shark skeletal muscle tropomyosin is a phosphoprotein
Hayley M, et al.
Journal of Muscle Research and Cell Motility, 29(2-5), 101-107 (2008)
Malik Shoaib Ahmad et al.
Proteins, 91(1), 22-31 (2022-08-05)
Bovine pancreatic trypsin was crystallized, in-complex with Lima bean trypsin inhibitor (LBTI) (Phaseolus lunatus L.), in the form of a ternary complex. LBTI is a Bowman-Birk-type bifunctional serine protease inhibitor, which has two independent inhibitory loops. Both of the loops

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service