Skip to Content
Merck
All Photos(1)

Key Documents

T1021

Sigma-Aldrich

Trypsin inhibitor

powder, suitable for isoelectric focusing (IEF)

Synonym(s):

SBTI

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Trypsin inhibitor from Glycine max (soybean), Isoelectric focusing marker, pI 4.6

biological source

Glycine max (soybean)

Quality Level

form

powder

mol wt

20,100 Da

technique(s)

isoelectric focusing (IEF): suitable

pI 

4.6

solubility

balanced salt solution: 1 mg/mL
concentrate: >10 mg/mL, hazy, amber-yellow
phosphate buffer: 10 mg/mL
water: 10 mg/mL
serum-free medium: soluble

shipped in

ambient

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

This inhibitor acts against trypsin, and chymotrypsin and plasmin to a lesser extent. It will also inhibit proteases with mechanisms similar to trypsin, plasma kallikrein and coagulation Factor X. The trypsin inhibitor will not act against metalloproteases, tissue-baseed kallikrein, acid proteases, or thio proteases. This inhibitor acts by forming a 1:1 stoichiometric complex with the protease active site, and then cleaving a single arginine-isoleucine bond on the inhibitor. The inhibition is both reversible and pH dependent.

Components

The soybean trypsin inhibitor is a monomeric protein containing 181 amino acid residues in a single polypeptide chain crosslinked by two disulfide bridges.

Unit Definition

One trypsin unit = A253 of 0.001 per minute with N-alpha-benzoyl-L-arginine ethyl ester (BAEE) as substrate at pH 7.6 at 25 °C.

Preparation Note

The trypsin inhibitor is soluble in water and phosphate buffers at 10 mg/mL. It is soluble in balanced salt solutions at 1 mg/mL and in serum-free media. Concentrated solutions greater than 10 mg/mL may be hazy and have a yellow to amber color. After trypsinizing cells, resuspend in 1 mL trypsin inhibitor solution at 1 mg/mL for every mL of trypsin solution used for dissociation. The cell suspension should then be centrifuged at 1000 rpm, forming a cell pellet.

Solutions can retain activity when stored short-term at 2-8° C. Solutions are stable in frozen aliquots at -20°C.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amaury Pereira-Acácio et al.
PloS one, 17(8), e0273385-e0273385 (2022-08-20)
We investigated the mechanisms by which chronic administration of a multideficient diet after weaning alters bodily Na+ handling, and culminates in high systolic blood pressure (SBP) at a juvenile age. From 28 to 92 days of age, weaned male Wistar
Kine Gregersen et al.
International journal of general medicine, 4, 555-560 (2011-09-03)
Food hypersensitivity is commonly suspected, but seldom verified. Patients with subjective food hypersensitivity suffer from both intestinal and extraintestinal health complaints. Abnormalities of the enterochromaffin cells may play a role in the pathogenesis. The aim of this study was to
Astrid F Nottebaum et al.
The Journal of experimental medicine, 205(12), 2929-2945 (2008-11-19)
We have shown recently that vascular endothelial protein tyrosine phosphatase (VE-PTP), an endothelial-specific membrane protein, associates with vascular endothelial (VE)-cadherin and enhances VE-cadherin function in transfected cells (Nawroth, R., G. Poell, A. Ranft, U. Samulowitz, G. Fachinger, M. Golding, D.T.
Shingo Kikuchi et al.
The Plant cell, 21(6), 1781-1797 (2009-06-18)
Chloroplast protein import is mediated by two hetero-oligomeric protein complexes, the Tic and Toc translocons, which are located in the inner and outer envelope membranes. At the inner membrane, many Tic components have been identified and characterized, but it remains
Eunice Andrè et al.
The Journal of clinical investigation, 118(7), 2574-2582 (2008-06-24)
Cigarette smoke (CS) inhalation causes an early inflammatory response in rodent airways by stimulating capsaicin-sensitive sensory neurons that express transient receptor potential cation channel, subfamily V, member 1 (TRPV1) through an unknown mechanism that does not involve TRPV1. We hypothesized

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What is the molecular weight and amino acid sequence of trypsin inhibitor from Glycine max (soybean)?

    1 answer
    1. Soybean trypsin inhibitor is a mixture of the three types. We have not characterized our trypsin inhibitor products in terms of how much of each of the three types may be present. All three have a molecular weight of between 20,000 and 20,200 daltons, so one might use 20,100 daltons as an average.The three types vary in the amino acids at a few positions. The detailed summary of the three types (or variants) was published in the following linked reference:     J. Biochem. (Japan), 98, 435-448 (1985).Note that the authors use Tia, Tib and Tic to indicate the three variants.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service