Skip to Content
Merck
All Photos(1)

Key Documents

T2580

Sigma-Aldrich

Trapoxin A

≥98% (HPLC), from Helicoma ambiens

Synonym(s):

Cyclo((S)-phenylalanyl-(S)-phenylalanyl-(R)-pipecolinyl-(2S,9S)-2-amino-8-oxo-9,10-epoxydecanoyl), RF 1023A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H42N4O6
CAS Number:
Molecular Weight:
602.72
UNSPSC Code:
12352200
NACRES:
NA.77

biological source

Helicoma ambiens

Quality Level

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: soluble 0.9-1.10 mg/ml, clear, colorless to faintly yellow (may be further diluted 20 fold in H2O)
chloroform: soluble
methanol: soluble

shipped in

dry ice

storage temp.

−20°C

SMILES string

N21[C@H](CCCC2)C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C1=O)Cc5ccccc5)Cc4ccccc4)CCCCCC(=O)[C@H]3OC3

InChI

1S/C34H42N4O6/c39-29(30-22-44-30)18-9-3-8-16-25-31(40)36-26(20-23-12-4-1-5-13-23)32(41)37-27(21-24-14-6-2-7-15-24)34(43)38-19-11-10-17-28(38)33(42)35-25/h1-2,4-7,12-15,25-28,30H,3,8-11,16-22H2,(H,35,42)(H,36,40)(H,37,41)/t25-,26-,27-,28+,30-/m0/s1

InChI key

GXVXXETYXSPSOA-UFEOFEBPSA-N

Application

Trapoxin A has been used:
  • to study its effects on the inhibition of histone deacetylase 11 (HDAC11)
  • to study its effects on the inhibition of HDAC3 in human cell lines
  • to study its effects on the inhibition of HDAC6 in rat pyramidal neurons

Biochem/physiol Actions

Trapoxin A is a cyclotetrapeptide and a histone deacetylase (HDAC) inhibitor. It increases the level of chromatin acetylation associated with histone H3 at low nanomolar concentrations. Unlike the reversible HDAC inhibition induced by TCA, Trapoxin A irreversibly inhibites HDAC activity in crude cell lysates, and induces the accumulation of hyperacetylated core histones in a number of mammalian cell lines and tissues. Histone acetylation and methylation have been studied extensively for their anti-tumor activities in carcinogenesis and Trapoxin has been suggested as a potential anticancer agent for pre-clinical trials.
Trapoxin is a cyclotetrapeptide and a histone deacetylase (HDAC) inhibitor.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Preparation Note

Trapoxin A is soluble in DMSO at a concentration ranging from 0.9-1.10 mg/ml and yields a clear, colorless to faint yellow solution. It is also soluble in chloroform and methanol.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John Nielsen
Current opinion in chemical biology, 6(3), 297-305 (2002-05-23)
Combinatorial syntheses allow production of compound libraries in an expeditious and organized manner immediately applicable for high-throughput screening. Natural products possess a pedigree to justify quality and appreciation in drug discovery and development. Currently, we are seeing a rapid increase
R Furumai et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(1), 87-92 (2001-01-03)
Trichostatin A (TSA) and trapoxin (TPX) are potent inhibitors of histone deacetylases (HDACs). TSA is proposed to block the catalytic reaction by chelating a zinc ion in the active-site pocket through its hydroxamic acid group. On the other hand, the
[A new target of cancer therapy: advances in the study of histone deacetylase].
Ai-Lin Liu et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 40(7), 585-590 (2005-10-04)
P L Sheridan et al.
Genes & development, 11(24), 3327-3340 (1998-02-07)
Specific inhibitors of histone deacetylase, such as trichostatin A (TSA) and trapoxin (TPX), are potent inducers of HIV-1 transcription in latently infected T-cell lines. Activation of the integrated HIV-1 promoter is accompanied by the loss or rearrangement of a positioned
Hua Cao et al.
Blood, 103(2), 701-709 (2003-08-16)
We investigated the induction of human gamma globin gene activity by 3 classes of histone deacetylase inhibitors: amide analogues of trichostatin A, hydroxamic acid analogues of trapoxin, and scriptaid and its analogues. The screening consisted of measuring the effects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service