Skip to Content
Merck
All Photos(3)

Documents

T6543

Sigma-Aldrich

Tricosanoic acid

≥99% (capillary GC)

Synonym(s):

Tricosylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)21COOH
CAS Number:
Molecular Weight:
354.61
Beilstein:
1795192
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥99% (capillary GC)

form

powder

mp

77-79 °C (lit.)

functional group

carboxylic acid

lipid type

saturated FAs

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C23H46O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23(24)25/h2-22H2,1H3,(H,24,25)

InChI key

XEZVDURJDFGERA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Kolusheva et al.
Journal of the American Chemical Society, 123(3), 417-422 (2001-07-18)
Biomolecular recognition of antigens and epitopes by antibodies is a fundamental event in the initiation of immune response and plays a central role in a variety of biochemical processes. Peptide binding requires, in many cases, presentation of the peptides at
Takahisa Mori et al.
Nutrients, 12(8) (2020-08-19)
Dietary triglycerides influence fatty acid (FA) serum concentrations and weight percentages (wt %), which may be associated with the age of onset of acute ischemic stroke (AIS). We investigated the correlations between serum FA levels and proportions at admission and
N C Shantha et al.
Journal of chromatography, 533, 1-10 (1990-11-30)
Tricosanoic acid (23:0) and cis-15-tetracosenoic acid (nervonic acid, 24:1 n-9) were compared as choices suitable for use as internal standards in the quantitation of eicosapentaenoic (EPA) and docosahexaenoic (DHA) fatty acids. Experiments conducted included: (a) comparison of the flame ionisation
Gemma C Shearman et al.
Langmuir : the ACS journal of surfaces and colloids, 23(13), 7276-7285 (2007-05-17)
Inverse bicontinuous cubic lyotropic phases are a complex solution to the dilemma faced by all self-assembled water-amphiphile systems: how to satisfy the incompatible requirements for uniform interfacial curvature and uniform molecular packing. The solution reached in this case is for
Felipe Pinheiro de Souza et al.
PloS one, 15(1), e0226977-e0226977 (2020-01-03)
Nutritional improvements in intensive aquaculture production systems is necessary for the reduction of stress, maximum utilization of nutritional components, and expression of the genetic potential of fish. The objective of this study was to evaluate the hemato-immunological, and histological parameters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service