Skip to Content
Merck
All Photos(1)

Key Documents

H5149

Supelco

Heneicosanoic acid

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)19COOH
CAS Number:
Molecular Weight:
326.56
Beilstein:
1711889
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

plant

Quality Level

grade

analytical standard

Assay

≥99% (capillary GC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

74-75 °C (lit.)

application(s)

food and beverages

format

neat

functional group

carboxylic acid

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C21H42O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21(22)23/h2-20H2,1H3,(H,22,23)

InChI key

CKDDRHZIAZRDBW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Heneicosanoic acid has been used as an internal standard in the analysis of fatty acid and conjugated linoleic acid isomers in meat samples using gas chromatography (GC) technique.
Heneicosanoic acid is used as an internal standard to quantify the methyl esters in fat extract from animal samples using chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haisu Shi et al.
Microbial cell factories, 15(1), 117-117 (2016-07-02)
Delta-6 desaturase (FADS6) is a key bifunctional enzyme desaturating linoleic acid (LA) or α-linolenic acid (ALA) in the biosynthesis of polyunsaturated fatty acids (PUFAs). In previous work, we analyzed the substrate specificity of two FADS6 enzymes from Mortierella alpina ATCC
Effect of microwave cooking or broiling on selected nutrient contents, fatty acid patterns and true retention values in separable lean from lamb rib-loins, with emphasis on conjugated linoleic acid
Maranesi.M, et al.
Food Chemistry, 90(1-2), 207-218 (2005)
Antonio Molino et al.
International journal of environmental research and public health, 15(11) (2018-11-06)
The exploration of new food sources and natural products is the result of the increase in world population as well as the need for a healthier diet; in this context, microalgae are undoubtedly an interesting solution. With the intent to
Ronan Lordan et al.
Foods (Basel, Switzerland), 8(5) (2019-05-30)
The in vitro antithrombotic properties of polar lipid constituents of malted grain (MG), pelleted hops (PH), brewer's spent grain (BSG), spent hops (SH), wort, and bottled beer from the same production line were assessed in human platelets. The total lipids
Alexandre G Torres et al.
The British journal of nutrition, 95(3), 517-524 (2006-03-04)
Maternal adipose tissue is a major contributor to breast milk long-chain fatty acids, probably through the pool of plasma NEFA. The fatty acid composition of the erythrocyte membrane (EM) is a biochemical index of the intake of fatty acids not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service