71015BC
Assurance® GDS MPX Top 7 STEC Kit
suitable for PCR testing in Food and Beverages applications
About This Item
Recommended Products
product name
GDS MPX Top 7 STEC, BioControl, Molecular based PCR test for the detection of E. coli O157:H7 and the "Top Six" non-O157 Shiga Toxigenic E. coli (STEC) in food
Agency
AOAC® (PTM)
Quality Level
shelf life
6‑12 mo.
packaging
pkg of 100 tests
manufacturer/tradename
Assurance GDS®
BioControl Systems 71015-100
BioControl Systems 71015-576
technique(s)
PCR: suitable
application(s)
food and beverages
pathogen testing
compatibility
for use with GDS Rotor-Gene
storage temp.
2-8°C
suitability
enterovirulent E. coli (EEC) (shiga toxigenic)
General description
Application
Features and Benefits
- Following sample preparation with IMS against the Top 7 O groups, samples are analyzed with a single multiplex assay for the presence of (eae), stx1, and stx2
- Assurance® GDS MPX Top 7 STEC assay includes a separate target and differentiated result for E. coli O157:H7
- Single-step 10-18 h enrichment in mEHEC®, enrichment media common to all BioControl methods for E.coli O157:H7, Shiga Toxin genes, and Top STEC
Components
- MPX Top STEC Amplification Tubes
- Top 7 STEC Concentration ReagentResuspension Buffer TqTop STEC Wash Solution
Compatibility
Testing Method
Legal Information
configured for
required but not provided
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service