Skip to Content
Merck
All Photos(2)

Documents

T5076

Sigma-Aldrich

Anti-β-Tubulin Isotype III antibody, Mouse monoclonal

clone SDL.3D10, purified from hybridoma cell culture

Synonym(s):

Anti-CDCBM, Anti-CDCBM1, Anti-CFEOM3, Anti-CFEOM3A, Anti-FEOM3, Anti-TUBB4, Anti-beta-4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

SDL.3D10, monoclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human, boar, bovine

packaging

antibody small pack of 25 μL

concentration

~2 mg/mL

technique(s)

dot blot: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
microarray: suitable
western blot: 0.1-0.2 μg/mL using rat brain extract

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TUBB3(10381)
mouse ... Tubb3(22152)
rat ... Tubb3(246118)

General description

Tubulin β 3 class III (TUBB3) encodes for the microtubule protein tubulin β‐3, which is majorly expressed in neurons. The gene is located on human chromosome 16q24.3.
Tubulin is the main constituent of microtubules made up of two similar polypeptides α and β, each with 50 kD (approx.) molecular weight. It has several cellular functions like mitosis, meiosis, some forms of organellar movement, intracellular transport, flagellar movement and cytoskeletal functions. Monoclonal anti-β-tubulin isotype III antibody can be used in microarray and western blotting. It can also be used in immunoblotting. Mouse anti-β-tubulin isotype III antibody reacts specifically with an epitope present on the β-tubulin (isotype III) of bovine, rat, mouse, boar and human but not with tubulin present in chicken brain preparation or in a chicken fibroblasts cell line.

Immunogen

peptide corresponding to the carboxy-terminal sequence of human β-tubulin isotype III, conjugated to BSA.

Application

Monoclonal Anti-β-Tubulin Isotype III antibody produced in mouse has been used in immunolabelling and flow cytometry.
Monoclonal anti-β-tubulin isotype III antibody (diluted 1:500) can be used as a primary antibody in immunocytochemical analysis of neurons. It can also be used for the localization of isotype III of β-tubulin by several immunochemical assays like dot blot, ELISA and immunohistochemistry.

Biochem/physiol Actions

Tubulin β 3 class III (TUBB3) regulates neurogenesis and is involved in axon guidance and maintenance. Mutations in this gene is associated with hypoplasia of commissural axon (CA), congenital fibrosis of the extraocular muscles type 3 (CFEOM3) and dysgyria.

Physical form

Solution in 0.01 M phosphate buffered saline containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Childhood-onset schizophrenia case with 2.2 Mb deletion at chromosome 3p12. 2-p12. 1 and two large chromosomal abnormalities at 16q22. 3-q24. 3 and Xq23-q28
Rudd D, et al.
Clinical case reports, 3(4), 201-207 (2015)
Two unique TUBB3 mutations cause both CFEOM3 and malformations of cortical development
Whitman MC, et al.
American Journal of Medical Genetics. Part A, 170(2), 297-305 (2016)
Evaluation of tubulin beta-3 as a novel senescence-associated gene in melanocytic malignant transformation
Orfanidis K, et al.
Pigment Cell & Melanoma Research, 30(2), 243-254 (2017)
Cytoskeleton in action: lissencephaly, a neuronal migration disorder
Moon HM and Wynshaw-Boris A
Wiley interdisciplinary reviews. Developmental biology, 2(2), 229-245 (2013)
J Pĕknicová et al.
Biology of reproduction, 65(3), 672-679 (2001-08-22)
The exposure of tubulin epitopes was studied in ejaculated boar spermatozoa using a panel of four monoclonal antibodies specific to the N-terminal or C-terminal structural domains of tubulin and three monoclonal antibodies against class III beta-tubulin. The specificity of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service