Skip to Content
Merck
All Photos(3)

Documents

N5413

Sigma-Aldrich

Anti-Nestin antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-FJ21841, Anti-NES, Anti-Nbla00170

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~110 kDa (cleaved)
antigen ~177 kDa
antigen ~270 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

flow cytometry: 1:200-1:500
immunohistochemistry: 1:100-1:200
western blot: 1:500-1:1,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NES(10763)
mouse ... Nes(18008)
rat ... Nes(25491)

General description

Nestin (NES) is a class VI cytoplasmic protein. It is expressed in the intermediate filaments and has a molecular weight of 177kDa. The gene encoding it is localized on human chromosome 1q23.1 with four exons.

Immunogen

synthetic peptide corresponding to residues 254-270 of human Nestin. This sequence is 93% identical in rat.

Biochem/physiol Actions

Nestin (NES) functions as a regulator of proteins which are involved in cell growth. It also interacts with other proteins of the intermediate filament. NES has been studied as a marker of neural stem cells.

Physical form

solution in phosphate buffered saline, containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nianping Zhang et al.
International journal of molecular medicine, 40(3), 814-824 (2017-07-18)
Pluripotent stem cells (PSCs) are regarded as potential sources that provide specific neural cells for cell therapy in some nervous system diseases. However, the mechanisms underlying the neural differentiation of PSCs remain largely unknown. MicroRNAs (miRNAs or miRs) are a class of
Y Chen et al.
Osteoarthritis and cartilage, 23(10), 1728-1735 (2015-06-02)
Osteoarthritis (OA) is a progressive joint disorder. To date, there is not effective medical therapy. Joint distraction has given us hope for slowing down the OA progression. In this study, we investigated the benefits of joint distraction in OA rat
Chhagan Bihari et al.
Hepatology (Baltimore, Md.), 64(4), 1273-1288 (2016-08-04)
Bone marrow (BM) is a reservoir for immune and hematopoietic cells and critical for tissue repair and regeneration. All of these functions are severely altered in cirrhosis. We investigated the cellular and functional state of BM in cirrhosis patients. We
Mohamed Ahmed Eladl et al.
International journal of nephrology and renovascular disease, 10, 61-76 (2017-03-07)
Several renal disorders affect the glomerular podocytes. Compensatory structural and functional changes have been observed in animals that have undergone unilateral renal ablation. These changes occur as a pliant response to quench the increased functional demand to maintain homeostasis of
Wei Zhao et al.
Scientific reports, 6, 21176-21176 (2016-02-18)
Restenosis or occlusion after vascular procedures is ascribed to intimal hyperplasia. Transforming growth factor (TGF)-β1 is involved in recruitment of mesenchymal stem cells (MSCs) following arterial injury, and its release from latent TGF-binding protein by matrix metalloproteinase (MMP)-14-induced proteolysis contributes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service