Skip to Content
Merck
All Photos(3)

Key Documents

T4632

Sigma-Aldrich

Glyceryl trinonadecanoate

≥99%

Synonym(s):

TG(19:0/19:0/19:0), Trinonadecanoin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H116O6
CAS Number:
Molecular Weight:
933.56
MDL number:
UNSPSC Code:
41141833
PubChem Substance ID:
NACRES:
NA.25

biological source

plant

Assay

≥99%

form

powder

technique(s)

GC/GC: suitable

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCCC

InChI

1S/C60H116O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-52-58(61)64-55-57(66-60(63)54-51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)56-65-59(62)53-50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h57H,4-56H2,1-3H3

InChI key

IZTDYXKTXNJHMQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adriano Francescangeli et al.
Journal of AOAC International, 103(2), 449-455 (2019-09-20)
Adulteration of diesel fuel by the addition of vegetable oil is a problem that touches several countries around the world and bypasses the complexity of the specifications regarding the automotive diesel fuel distinguishing between customs, fiscal, and commercial-environmental specifications. At
I M Remmers et al.
Bioresource technology, 244(Pt 2), 1384-1392 (2017-05-10)
This work provides a novel quantitative comparison of batch versus continuous microalgal lipid production in the wild type and starchless mutant strain of Acutodesmus obliquus. Both strains showed higher TAG yields on light under batch operation compared to continuous nitrogen
Dirk Volker Woortman et al.
Frontiers in bioengineering and biotechnology, 7, 481-481 (2020-02-11)
A multitude of human nutritional supplements based on Chlorella vulgaris biomass has recently been introduced to the specialty food market. In this study, an analysis of total folate contents in Chlorella sp. and a series of marine microalgae was conducted
A Lohninger et al.
Analytical biochemistry, 171(2), 366-372 (1988-06-01)
An accurate capillary gas-chromatographic method with trinonadecanoylglycerol as internal standard for determining triacylglycerols in human serum and other biological sources is described. After serum extraction, total triacylglycerol and triacylglycerol species (differing in the number of carbon atoms in the acyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service