Skip to Content
Merck
All Photos(1)

Documents

394866

Supelco

N-Methyl-N-(trimethylsilyl)trifluoroacetamide

synthesis grade

Synonym(s):

N-Trimethylsilyl-N-methyl trifluoroacetamide, MSTFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CON(CH3)Si(CH3)3
CAS Number:
Molecular Weight:
199.25
Beilstein:
1941550
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization
synthesis grade

vapor density

>1 (vs air)

vapor pressure

8.8 mmHg ( 27 °C)

Assay

≥98%

form

liquid

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

refractive index

n20/D 1.38 (lit.)

bp

130-132 °C (lit.)

density

1.075 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CN(C(=O)C(F)(F)F)[Si](C)(C)C

InChI

1S/C6H12F3NOSi/c1-10(12(2,3)4)5(11)6(7,8)9/h1-4H3

InChI key

MSPCIZMDDUQPGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Methyl-N-(trimethylsilyl)trifluoroacetamide (MSTFA) is a derivatizing reagent. It can be used as alternatives to diazomethane.

Application

Silylating agent used to form volatile derivatives for GC-MS analysis.
Improved silylation procedure for simultaneous detection of steroid hormones 17-α-ethynylestradiol and estrone.

Other Notes

Reagent for trimethylsilyl.

related product

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Tina Kosjek et al.
Environment international, 31(5), 679-685 (2005-05-25)
Pharmacologically active substances used to treat human and animal illnesses can enter the aquatic environment via effluents from wastewater treatment plants or in the case of veterinary drugs directly through liquid manure discharge. Some of these substances enter the environment
Stina K Lien et al.
Journal of chromatography. A, 1247, 118-124 (2012-06-15)
GC-MS analysis of silylated metabolites is a sensitive method that covers important metabolite groups such as sugars, amino acids and non-amino organic acids, and it has become one of the most important analytical methods for exploring the metabolome. Absolute quantitative
Hee-Kyung Jeon et al.
Journal of chromatography. A, 1131(1-2), 192-202 (2006-08-08)
A novel method has been developed to simultaneously determine and quantify seven organic UV filters employing liquid (solid)-liquid extraction, derivatization with N-methyl-N-(trimethylsilyl) trifluoroacetamide (MSTFA) and gas chromatography with mass spectrometric detection in various environmental matrices. The UV filters determined were:
Iria González-Mariño et al.
Journal of chromatography. A, 1217(11), 1748-1760 (2010-02-13)
An alternative method for the sensitive determination of several drugs of abuse and some of their metabolites in surface and sewage water samples is proposed. Analytes are concentrated using a solid-phase extraction (SPE) sorbent, converted into the corresponding trimethylsilyl derivatives
L Damasceno et al.
Journal of mass spectrometry : JMS, 35(11), 1285-1294 (2000-12-13)
An evaluation of derivatization procedures for the detection of beta(2)-agonists is presented. The study was performed with the beta(2)-agonists bambuterol, clenbuterol, fenoterol, formoterol, salbutamol, salmeterol and terbutaline. Different derivatizating agents were employed, aiming to obtain derivatives with high selectivity to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service