Skip to Content
Merck
All Photos(3)

Key Documents

T3882

Sigma-Aldrich

Glyceryl tritridecanoate

≥99%

Synonym(s):

TG(13:0/13:0/13:0), 1,2,3-Tritridecanoylglycerol, Tritridecanoin (C13:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H80O6
CAS Number:
Molecular Weight:
681.08
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Quality Level

Assay

≥99%

form

powder

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCC)OC(=O)CCCCCCCCCCCC

InChI

1S/C42H80O6/c1-4-7-10-13-16-19-22-25-28-31-34-40(43)46-37-39(48-42(45)36-33-30-27-24-21-18-15-12-9-6-3)38-47-41(44)35-32-29-26-23-20-17-14-11-8-5-2/h39H,4-38H2,1-3H3

InChI key

UDXANBFMQUOKTQ-UHFFFAOYSA-N

Related Categories

Application

Glyceryl tritridecanoate has been used as an internal standard for the extraction of the total lipids from the muscle homogenate. It has also been used in toluene as an internal standard to assess the lipid content and fatty acid (FA) composition of forages and concentrates using a one-step extraction-transesterification procedure.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luís M Rodríguez-Alcalá et al.
Journal of agricultural and food chemistry, 65(35), 7680-7686 (2017-08-10)
This study aimed to assess the oral absorption and plasma kinetics of two main isomers contained in commercial conjugated linoleic acid (CLA)-rich oil (Tonalin TG-80), rumenic acid (RA), and C18:2 trans-10, cis-12. The isomer plasma disposition after the single oral
Ana Luiza Fontes et al.
Scientific reports, 8(1), 15591-15591 (2018-10-24)
Current research on lipids is highlighting their relevant role in metabolic/signaling pathways. Conjugated fatty acids (CFA), namely isomers of linoleic and linolenic acid (i.e. CLA and CLNA, respectively) can positively modulate inflammation processes and energy metabolism, promoting anti-carcinogenic and antioxidant
Jimena Lavandera et al.
The British journal of nutrition, 118(11), 906-913 (2017-11-28)
Conjugated linoleic acid (CLA) might regulate the lipid depots in liver and adipose tissue. As there is an association between maternal nutrition, fat depots and risk of offspring chronic disease, the aim was to investigate the effect of maternal CLA
Laura Goracci et al.
Chemical research in toxicology, 33(1), 258-270 (2019-12-11)
The importance of adsorption, distribution, metabolism, excretion, and toxicity (ADMET) analysis is expected to grow substantially due to recent failures in detecting severe toxicity issues of new chemical entities during preclinical/clinical development. Traditionally, safety risk assessment studies for humans have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service