T2193
Greiner centrifuge tubes
capacity 50 mL, conical bottom (with skirt), diam. × L 30 mm × 115 mm
Synonym(s):
centrifuge tubes, culture tubes, plastic centrifuge tube, plastic culture tube, Greiner centrifuge tubes
About This Item
Recommended Products
material
cap
conical bottom (with skirt)
polypropylene tube
sterility
sterile
feature
RCF 3,200 × g
graduations
packaging
case of 300 (6 x packs of 25EA)
manufacturer/tradename
Greiner 210270
capacity
50 mL
diam. × L
30 mm × 115 mm
color
blue cap
colorless tube
Looking for similar products? Visit Product Comparison Guide
General description
High quality, crystal-clear polypropylene centrifuge tubes. Centrifuge tubes can be safely centrifuged to approximately 3,200 × g (50 mL size) and 3,500 × g (15 mL size) depending on the specific gravity of the solution being centrifuged and the type of rotor.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service