Skip to Content
Merck
All Photos(2)

Key Documents

SAB5600206

Sigma-Aldrich

Anti-alpha-Tubulin antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM113, purified immunoglobulin

Synonym(s):

Tubulin alpha-1A chain, Tubulin alpha-1B chain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

recombinant

expressed in HEK 293 cells

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM113, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

all

technique(s)

ChIP: 1:200
ELISA: suitable (assay dependent concentration)
flow cytometry: suitable (assay dependent concentration)
immunocytochemistry: 1:200
immunohistochemistry: 1:200
immunoprecipitation (IP): 1:200
western blot: 1:1000

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TUBA1A(7846)

Related Categories

Specificity

This antibody reacts to alpha-Tubulin, including Tubulin alpha-1A chain and Tubulin alpha-1B chain.

Immunogen

A peptide corresponding to the C-terminus of alpha-Tubulin

Physical form

Solution in phosphate buffered saline containing 50% Glycerol/PBS with 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fei Shen et al.
Oncology letters, 20(5), 223-223 (2020-09-25)
An increasing amount of evidence has demonstrated the importance of microRNAs (miRNAs/miRs) in the tumorigenesis of malignant types of cancer, and data retrieved from The Cancer Genome Atlas database revealed that miR-3690 was upregulated in thyroid cancer (TC). The present
Milaid Granadillo Rodríguez et al.
Frontiers in genome editing, 5, 1196697-1196697 (2023-06-16)
APOBEC3 (A3) enzymes deaminate cytosine to uracil in viral single-stranded DNA as a mutagenic barrier for some viruses. A3-induced deaminations can also occur in human genomes resulting in an endogenous source of somatic mutations in multiple cancers. However, the roles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service