T3405
3,3′,5,5′-Tetramethylbenzidine dihydrochloride
chromogenic, tablet
Synonym(s):
4,4′-Diamino-3,3′,5,5′-tetramethylbiphenyl dihydrochloride
About This Item
Recommended Products
product name
3,3′,5,5′-Tetramethylbenzidine dihydrochloride, tablet, 1 mg substrate per tablet
form
tablet
mp
≥300 °C
storage temp.
2-8°C
SMILES string
Cl[H].Cl[H].Cc1cc(cc(C)c1N)-c2cc(C)c(N)c(C)c2
InChI
1S/C16H20N2.2ClH/c1-9-5-13(6-10(2)15(9)17)14-7-11(3)16(18)12(4)8-14;;/h5-8H,17-18H2,1-4H3;2*1H
InChI key
NYNRGZULARUZCC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- anti-mouse IgG (? specific) horseradish peroxidase-conjugated antibody in the ELISA of mice sera
- anti-mouse peroxidase antibody in the protein tyrosine phosphatase assay of naive and effector T cell lysate
- in enzymatic hydroperoxide (EH) assay and in prooxidants–antioxidants balance (PAB) assay of serum samples from diabetic patients
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service