Skip to Content
Merck
All Photos(1)

Key Documents

47784

Supelco

δ-Tocopherol

analytical standard

Synonym(s):

(+)-δ-Tocopherol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
E Number:
E309

grade

analytical standard

form

liquid

CofA

current certificate can be downloaded

feature

standard type fat soluble vitamin

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

-10 to -25°C

SMILES string

CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2cc(O)cc(C)c2O1

InChI

1S/C27H46O2/c1-20(2)10-7-11-21(3)12-8-13-22(4)14-9-16-27(6)17-15-24-19-25(28)18-23(5)26(24)29-27/h18-22,28H,7-17H2,1-6H3/t21-,22-,27-/m1/s1

InChI key

GZIFEOYASATJEH-VHFRWLAGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

δ-Tocopherol, a homolog of vitamin E, is a phytochemical with potent antioxidant activity. It has been investigated for its potential medical, biological, and physio-chemical applications.

Application

δ-Tocopherol may be used as an analytical reference standard for the determination of the analyte in food and beverage products and crops by high-performance liquid chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromatographic separation of tocopherols
Pyka A and Sliwiok J
Journal of Chromatography A, 935(1-2), 71-76 (2001)
A small-scale sample preparation method with HPLC analysis for determination of tocopherols and tocotrienols in cereals
Ryynanen M, et al.
Journal of food composition and analysis, 17(6), 749-765 (2004)
A rapid procedure for analysing rice bran tocopherol, tocotrienol and gamma-oryzanol contents
Chen MH and Bergman CJ
Journal of food composition and analysis, 18(2-3), 139-151 (2005)
Determination of vitamins E (alpha, gamma-and delta-tocopherol) and D (cholecalciferol and ergocalciferol) by liquid chromatography in milk, fruit juice and vegetable beverage
Barba FJ, et al.
European Food Research and Technology, 232(5), 829-836 (2011)
Qing Jiang et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(51), 17825-17830 (2004-12-15)
gamma-Tocopherol (gammaT), the predominant form of vitamin E in diets, but not alpha-tocopherol, the major vitamin E form in tissues and supplements, inhibits proliferation of prostate cancer cells (LNCaP and PC-3) and lung cancer cells (A549). In contrast, at similar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service