Skip to Content
Merck
All Photos(1)

Key Documents

T1505

Sigma-Aldrich

Tetraisopropyl pyrophosphoramide

butyrylcholinesterase inhibitor

Synonym(s):

Tetra(monoisopropyl)pyrophosphortetramide, iso-OMPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H32N4O3P2
CAS Number:
Molecular Weight:
342.36
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

mp

149-151 °C

storage temp.

−20°C

SMILES string

CC(C)NP(=O)(NC(C)C)OP(=O)(NC(C)C)NC(C)C

InChI

1S/C12H32N4O3P2/c1-9(2)13-20(17,14-10(3)4)19-21(18,15-11(5)6)16-12(7)8/h9-12H,1-8H3,(H2,13,14,17)(H2,15,16,18)

InChI key

IOIMDJXKIMCMIG-UHFFFAOYSA-N

Gene Information

Application

Tetraisopropyl pyrophosphoramide has been used:
  • as a butyrylcholinesterase inhibitor to determine the proportions of butyrylcholinesterase (BChE) in cat and tiger plasma
  • to inhibit wild-type BChE in acetylcholinesterase assay
  • to selectively block the enzymatic activity of AChE

Biochem/physiol Actions

Selective inhibitor of butyrylcholinesterase

Warning

WARNING: Extremely hazardous! Be aware of the risk and familiar with safety procedures before you use this product.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Resistance to organophosphorus agent toxicity in transgenic mice expressing the G117H mutant of human butyrylcholinesterase
Wang YW, et al.
Toxicology and Applied Pharmacology, 196(3), 356-366 (2004)
Souren Mkrtchian et al.
Experimental physiology, 105(9), 1634-1647 (2020-07-12)
What is the central question of this study? Are carotid bodies (CBs) modulated by the damage-associated molecular patterns (DAMPs) and humoral factors of aseptic tissue injury? What are the main findings and their importance? DAMPs (HMGB1, S100 A8/A9) and blood
Postnatal development of nitrergic and cholinergic structures in rat spinal cord
Bolekova A, et al.
Archives Italiennes de Biologie, 149(3), 293-302 (2011)
Liyi Geng et al.
PloS one, 8(6), e67446-e67446 (2013-07-11)
Gene transfer of a human cocaine hydrolase (hCocH) derived from butyrylcholinesterase (BChE) by 5 mutations (A199S/F227A/S287G/A328W/Y332G) has shown promise in animal studies for treatment of cocaine addiction. To predict the physiological fate and immunogenicity of this enzyme in humans, a
Determination of the DNA sequences of acetylcholinesterase and butyrylcholinesterase from cat and demonstration of the existence of both in cat plasma.
Bartels CF, et al.
Biochemical Pharmacology, 60(4), 479-487 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service