Skip to Content
Merck
All Photos(2)

Key Documents

A7000

Sigma-Aldrich

Acetylcholine iodide

≥97%

Synonym(s):

ACh

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N(I)CH2CH2OCOCH3
CAS Number:
Molecular Weight:
273.11
Beilstein:
3571339
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥97%

form

powder

storage temp.

−20°C

SMILES string

[I-].CC(=O)OCC[N+](C)(C)C

InChI

1S/C7H16NO2.HI/c1-7(9)10-6-5-8(2,3)4;/h5-6H2,1-4H3;1H/q+1;/p-1

InChI key

SMBBQHHYSLHDHF-UHFFFAOYSA-M

Gene Information

human ... CHRM3(1131)

Looking for similar products? Visit Product Comparison Guide

Application

Acetylcholine is an endogenous neurotransmitter at cholinergic synapses that amplifies action potential of the sarcolemma thereby inducing muscle contractions. Acetylcholine iodide is used as an acetylcholine receptor agonist to identify, characterize and differentiate among types of cholinergic receptors. Acetylcholine iodide is used as a substrate to identify and characterize natural and mutated acetylcholinesterase(s).

Biochem/physiol Actions

Endogenous neurotransmitter at cholinergic synapses; amplifies action potential of the sarcolemma thereby inducing muscle contractions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iryna Andrusenko et al.
Angewandte Chemie (International ed. in English), 58(32), 10919-10922 (2019-06-19)
Orthocetamol is a regioisomer of the well-known pain medication paracetamol and a promising analgesic and an anti-arthritic medicament itself. However, orthocetamol cannot be grown as single crystals suitable for X-ray diffraction, so its crystal structure has remained a mystery for
Marie Briet et al.
Journal of the American Heart Association, 2(2), e000128-e000128 (2013-04-16)
Recent studies have raised concern about the safety of erythropoiesis-stimulating agents because of evidence of increased risk of hypertension and cardiovascular morbidity and mortality in chronic kidney disease (CKD) patients. In the present study, we investigated the effects of recombinant
Tracey Huynh et al.
Journal of medicinal chemistry, 56(20), 8196-8200 (2013-10-01)
The M4 mAChR is implicated in several CNS disorders and possesses an allosteric binding site for which ligands modulating the affinity and/or efficacy of ACh may be exploited for selective receptor targeting. We report the synthesis of a focused library
Xun Qian et al.
Arteriosclerosis, thrombosis, and vascular biology, 34(1), 127-135 (2013-11-02)
Intermediate and small conductance KCa channels IK1 (KCa3.1) and SK3 (KCa2.3) are primary targets of endothelial Ca(2+) signals in the arterial vasculature, and their ablation results in increased arterial tone and hypertension. Activation of IK1 channels by local Ca(2+) transients
Yingchuan B Qi et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(12), 5319-5325 (2013-03-22)
Excitatory acetylcholine motor neurons drive Caenorhabditis elegans locomotion. Coordinating the activation states of the backward-driving A and forward-driving B class motor neurons is critical for generating sinusoidal and directional locomotion. Here, we show by in vivo calcium imaging that expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service