Skip to Content
Merck
All Photos(3)

Key Documents

D6821

Sigma-Aldrich

Donepezil hydrochloride

≥98% (HPLC), powder, acetylcholinesterase inhibitor

Synonym(s):

1H-Inden-1-one, 2,3-dihydro-5,6-dimethoxy-2-[[1-(phenylmethyl)-4-piperidinyl]methyl]-, hydrochloride, E2020

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H29NO3 · HCl
CAS Number:
Molecular Weight:
415.95
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Donepezil hydrochloride, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

DMSO: ≥20 mg/mL
H2O: ≥20 mg/mL

storage temp.

room temp

SMILES string

O=C1C(CC2CCN(CC3=CC=CC=C3)CC2)CC4=CC(OC)=C(OC)C=C41.[H]Cl

InChI

1S/C24H29NO3.ClH.H2O/c1-27-22-14-19-13-20(24(26)21(19)15-23(22)28-2)12-17-8-10-25(11-9-17)16-18-6-4-3-5-7-18;;/h3-7,14-15,17,20H,8-13,16H2,1-2H3;1H;1H2

InChI key

HLJIZAKUNCTCQX-UHFFFAOYSA-N

Gene Information

human ... ACHE(43)

Looking for similar products? Visit Product Comparison Guide

Application

Donepezil hydrochloride has been used in dose-response experiment to study its efficacy in ameliorating nicotine withdrawal-induced deficits in contextual conditioning in mice. It has also been intraperitoneally injected into rats to study its ability to improve social behavioral deficits, repetitive behavior and hyperactivity.

Biochem/physiol Actions

Donepezil hydrochloride is a piperidine containing an organic compound and a non-competitive inhibitor. It is useful in treating Alzheimer′s disease, in which dementia is a prominent symptom. Donepezil is shown to induce cognitive ability and overall body function. It has a half-life of 70 hours.
Donepezil monohydrochloride monohydrate is a centrally acting reversible acetylcholinesterase inhibitor.
Donepezil monohydrochloride monohydrate is a reversible acetylcholinesterase inhibitor

Features and Benefits

This compound is featured on the Acetylcholine Synthesis and Metabolism page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haneul Kim et al.
Journal of ethnopharmacology, 211, 38-46 (2017-09-18)
The seeds of Ziziphus jujuba var. spinosa (Bunge) Hu ex H.F Chow (Rhamnaceae) and the roots of Codonopsis lanceolata (Siedbold & Zucc.) Benth. & Hook. f ex Trautv. (Campanulaceae), contained in the DHP1402, have long been used for treating dementia
Donepezil for dementia due to Alzheimer's disease
Birks Jacqueline S and Harvey Richard J
Cochrane database of systematic reviews (Online), 6 (2018)
Ji Hyun Kim et al.
Nutrients, 12(5) (2020-05-14)
Obesity increases risk of Alzheimer's Disease (AD). A high fat diet (HFD) can lead to amyloidosis and amyloid beta (Aβ) accumulation, which are hallmarks of AD. In this study, protective effects of the ethyl acetate fraction of Acer okamotoanum (EAO)
Sunhee Lee et al.
Phytotherapy research : PTR, 31(3), 425-432 (2017-02-07)
Acanthopanax koreanum Nakai (Araliaceae) is one of the most widely cultivated medicinal plants in Jeju Island, Korea, and the roots and stem bark of A. koreanum have been traditionally used as a tonic agent for general weakness. However, the use of
Gilbert Audira et al.
Biomolecules, 10(9) (2020-09-24)
Donepezil (DPZ) is an acetylcholinesterase inhibitor used for the clinical treatment of mild cognitive impairment. However, DPZ has been reported to have adverse effects, including causing abnormal cardiac rhythm, insomnia, vomiting, and muscle cramps. However, the existence of these effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service