Skip to Content
Merck
All Photos(4)

Key Documents

T0167

Sigma-Aldrich

D-(+)-Trehalose dihydrate

from Saccharomyces cerevisiae, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%

Synonym(s):

α,α-Trehalose, α-D-Glucopyranosyl-α-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · 2H2O
CAS Number:
Molecular Weight:
378.33
Beilstein:
5322018
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.75

Quality Level

biological source

Saccharomyces cerevisiae

product line

BioReagent

Assay

≥99%

form

powder

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

solubility

H2O: 50 mg/mL

SMILES string

[H]O[H].[H]O[H].OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11.2H2O/c13-1-3-5(15)7(17)9(19)11(21-3)23-12-10(20)8(18)6(16)4(2-14)22-12;;/h3-20H,1-2H2;2*1H2/t3-,4-,5-,6-,7+,8+,9-,10-,11-,12-;;/m1../s1

InChI key

DPVHGFAJLZWDOC-PVXXTIHASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trehalose is a non-reducing disaccharide, ubiquitously present in nature. It is used as a food preservative and also to retain enzymes, vaccines, cells and many other molecules. Trehalose is present as hemolymph sugar among the invertebrates.

Application

D-(+)-Trehalose dihydrate has been used in a slow-freezing method for freezing rabbit oocytes. It has also been used in solid-surface vitrification (SSV) of in vitro-matured oocytes. It is suitable for use as a cryoprotectant in a variety of cell freezing media.

Biochem/physiol Actions

Trehalose is essential for embryogenesis, carbon metabolism, photosynthesis and flowering in plants. It predominantly exists among the species that supports anhydrobiosis (life without water). It serves as a carbohydrate reserve in microorganisms and protects them from adverse conditions, such as heat, cold, dehydration, desiccation and oxidation. Trehalose acts as a chemical chaperone affecting protein folding. It is known to prevent amyloid generation and accummulation of β-amyloid in vitro. A similar effect of trehalose has been observed with huntingtin protein aggregation. Thus, it might serve as a protective agent in Alzheimer′s disease and Huntington′s disease.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trehalose metabolism: from osmoprotection to signaling
Iturriaga G, et al.
International Journal of Molecular Sciences, 10(9), 3793-3810 (2009)
Developmental competence of in vitro-fertilized porcine oocytes after in vitro maturation and solid surface vitrification: effect of cryopreservation on oocyte antioxidative system and cell cycle stage
Somfai T, et al.
Cryobiology, 55(2), 115-126 (2007)
Production of good-quality porcine blastocysts by in vitro fertilization of follicular oocytes vitrified at the germinal vesicle stage
Somfai T, et al.
Theriogenology, 73, 147-156 (2010)
Biochemical analysis of trehalose and its metabolizing enzymes in wheat under abiotic stress conditions.
El-Bashiti T, et al.
Plant Science, 169(1), 47-54 (2005)
Methodios Ximerakis et al.
Nature neuroscience, 22(10), 1696-1708 (2019-09-26)
The mammalian brain is complex, with multiple cell types performing a variety of diverse functions, but exactly how each cell type is affected in aging remains largely unknown. Here we performed a single-cell transcriptomic analysis of young and old mouse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service