Skip to Content
Merck
All Photos(1)

Key Documents

SRP6014

Sigma-Aldrich

HGF human

recombinant, expressed in CHO cells, ≥97% (SDS-PAGE), ≥97% (HPLC)

Synonym(s):

F-TCF, HGF, HGFB, Hepatopoietin (HPTA), Scatter Factor (SF)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in CHO cells

Assay

≥97% (HPLC)
≥97% (SDS-PAGE)

form

lyophilized

mol wt

80.017 kDa

packaging

pkg of 1 mg
pkg of 10 μg

technique(s)

cell culture | embryo: suitable

impurities

Endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... HGF(3082)

General description

Hepatocyte growth factor (HGF) is a multifunctional growth factor. HGF Human Recombinant produced in Chinese hamster ovary (CHO) is a heterodimer, non-glycosylated, polypeptide chain consisting of an α-chain of 463 amino acids and β-chain of 234 having a total molecular mass of 80.017kDa. The gene encoding this protein is localized on human chromosome 7q21.1. The HGF is purified by proprietary chromatographic techniques.

Biochem/physiol Actions

Hepatocyte growth factor (HGF) regulates both cell growth and cell motility. It exerts a strong mitogenic effect on hepatocytes and primary epithelial cells. HGF synergizes with interleukin-3 and granulocyte-macrophage colony-stimulating factor (GM-CSF) to stimulate colony formation of hematopoietic progenitor cells in vitro and may, therefore, also modulate hematopoiesis. It is expressed in brain tumors.

Physical form

The protein was lyophilized from a concentrated (1 mg/ml) solution containing PBS pH 7.4 containing 0.05% Tween 20 and 100 mM L-arginine.

Reconstitution

Centrifuge the vial prior to opening. It is recommended to reconstitute the lyophilized Hepatocyte Growth Factor (HGF) in sterile 18 MΩcm-1 H2O not less than 100 μg/mL, which can then be further diluted to other aqueous solutions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HGF: a multifunctional growth factor controlling cell scattering.
Stella MC and Comoglio PM
The International Journal of Biochemistry & Cell Biology, 31(12), 1357-1362 (1999)
Scatter factor/hepatocyte growth factor in brain tumor growth and angiogenesis.
Abounader R and Laterra J
Neuro-Oncology, 7(4), 436-451 (2005)
Synergistic effects of hepatocyte growth factor on human cord blood CD34+ progenitor cells are the result of c-met receptor expression.
Goff JP
Stem Cells, 14(5), 592-602 (1996)
Rajani Kanteti et al.
PloS one, 9(9), e105919-e105919 (2014-09-16)
Malignant pleural mesothelioma (MPM) is an aggressive disease with a poor prognosis. Studies have shown that both MET and its key downstream intracellular signaling partners, PI3K and mTOR, are overexpressed in MPM. Here we determined the combinatorial therapeutic efficacy of
Hiroshi Kusunoki et al.
Hypertension research : official journal of the Japanese Society of Hypertension, 37(6), 500-506 (2014-03-14)
The paracrine loop involving inflammatory cytokines between adipocytes and macrophages establishes a vicious cycle that aggravates pro-inflammatory changes in adipose tissue. The serum level of hepatocyte growth factor (HGF) is increased in metabolic syndrome, but whether HGF is beneficial or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service