Skip to Content
Merck
All Photos(1)

Key Documents

H5791

Sigma-Aldrich

Hepatocyte Growth Factor human

HumanKine®, ≥95% (SDS-PAGE), recombinant, expressed in HEK 293 cells, lyophilized powder, suitable for cell culture

Synonym(s):

HGF, Hepatopoietin A, Scatter Factor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Product Name

Hepatocyte Growth Factor human, HGF, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

≤20 ng/mL EC50

quality

endotoxin tested

mol wt

monomer 70 kDa (glycosylated)

packaging

pkg of 5x10 μg
pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

≤1 EU/mg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... HGF(3082)

Looking for similar products? Visit Product Comparison Guide

General description

The HGF gene spans ∼70 kb and consists of 18 exons interrupted by 17 introns. The organization of the human HGF gene is highly homologous to that of human plasminogen. HGF maps to the long arm of human chromosome 7, 7q21.1.

Biochem/physiol Actions

HGF is a multifunctional cytokine that promotes mitogenesis, migration, invasion, and morphogenesis. It is a potent mitogen, stimulating the growth of hepatocytes, renal tubular epithelial cells, epidermal keratinocytes, epidermal melanocytes, Mv1Lu (mink lung epithelial cells), and BALB/MK (mouse keratinocytes). HGF inhibits the growth of B6/F1 (mouse melanoma) cells, KB (human squamous carcinoma) cells, and HepG2 (human hepatoma) cells. HGF is the ligand for the tyrosine kinase receptor encoded by the met proto-oncogene.
Hepatocyte growth factor (HGF) is a potent mitogen for hepatocytes and a variety of cell types including endothelial and epithelial cells, melanocytes, and keratinocytes. It also mediates epithelial morphogenesis. HGF is the ligand for the tyrosine kinase receptor encoded by the met proto-oncogene.

Other Notes

The NSO-expressed recombinant HGF is greater than 95% heterodimeric HGF. The insect cell line-expressed recombinant HGF is a mixture of single chain HGF (predominant) with some heterodimeric HGF. Single chain HGF and heterodimeric HGF are equally active in in vitro assays.

Physical form

Lyophilized from a 0.2 μm filtered solution of 1x PBS.

Preparation Note

HumanKine HGF is expressed as a glycosylated 70 kDa single chain monomer in human HEK 293 cells. Production in human HEK 293 cells offers authentic glycosylation. Glycosylation contributes to stability in cell growth media and other applications.

Analysis Note

The specific activity was determined by the dose-dependent stimulation of the proliferation of the monkey epithelial cell line 4MBr-5.

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karène Mahtouk et al.
Biochimica et biophysica acta, 1806(2), 208-219 (2010-07-27)
Hepatocyte growth factor (HGF) and its receptor MET are essential during embryonic development and throughout postnatal life. However, aberrant MET activation, due to overexpression, mutations, or autocrine ligand production, contributes to the development and progression of a variety of human
T Nakamura et al.
Nature, 342(6248), 440-443 (1989-11-23)
Hepatocyte growth factor (HGF) is the most potent mitogen for mature parenchymal hepatocytes in primary culture, and seems to be a hepatotrophic factor that acts as a trigger for liver regeneration after partial hepatectomy and liver injury. The partial purification
Frieda Kage et al.
Journal of cell science, 135(15) (2022-08-17)
Cell migration frequently involves the formation of lamellipodia induced by Rac GTPases activating WAVE regulatory complex (WRC) to drive Arp2/3 complex-dependent actin assembly. Previous genome editing studies in B16-F1 melanoma cells solidified the view of an essential, linear pathway employing
Rajani Kanteti et al.
PloS one, 9(9), e105919-e105919 (2014-09-16)
Malignant pleural mesothelioma (MPM) is an aggressive disease with a poor prognosis. Studies have shown that both MET and its key downstream intracellular signaling partners, PI3K and mTOR, are overexpressed in MPM. Here we determined the combinatorial therapeutic efficacy of
Hiroshi Kusunoki et al.
Hypertension research : official journal of the Japanese Society of Hypertension, 37(6), 500-506 (2014-03-14)
The paracrine loop involving inflammatory cytokines between adipocytes and macrophages establishes a vicious cycle that aggravates pro-inflammatory changes in adipose tissue. The serum level of hepatocyte growth factor (HGF) is increased in metabolic syndrome, but whether HGF is beneficial or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service