Skip to Content
Merck
All Photos(1)

Key Documents

E4127

Sigma-Aldrich

Epidermal Growth Factor from murine submaxillary gland

lyophilized, suitable for cell culture

Synonym(s):

EGF, Epidermal cell growth factor

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Product Name

Epidermal Growth Factor from murine submaxillary gland, EGF, suitable for cell culture

biological source

mouse submaxillary glands

Quality Level

form

lyophilized powder

potency

0.1-10 ng/mL ED50/EC50

quality

endotoxin tested

mol wt

6 kDa

packaging

pkg of 0.1 and 5X0.1 mg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

≤50 EU/vial (LAL test)

color

white to off-white

solubility

water: soluble, clear, colorless (1 vial is reconstituted with 0.5 mL of 0.85% NaCl)

UniProt accession no.

storage temp.

2-8°C

Gene Information

mouse ... Egf(13645)

Looking for similar products? Visit Product Comparison Guide

Application

Epidermal Growth Factor (EGF) from murine submaxillary gland has been used as a mitogen, triggering the cell to commence mitosis, in a variety of cell lines. In tissue cultures, EGF acts to reduce or eliminate the requirement for serum and can be used in conjunction with other media additives and hormone. The working concentration for EGF is generally 0.1-10 ng/mL.

Biochem/physiol Actions

EGF is mitogenic for a variety of epidermal and epithelial cells, including fibroblasts, glial cells, vascular and corneal endothelial cells, bovine granulosa, HeLa cells, SV40-3T3 cells and mammary epithelial cells, rabbit chondrocytes. EGF also plays a biological role in inhibition of gastric acid secretion, support of growth during fetal development and neuromodulation in the central nervous system. Stimulation of ion fluxes, glucose transport, glycolysis, and synthesis of DNA, RNA and proteins are all cellular metabolic effects of EGF.

Components

Epidermal Growth Factor (EGF) is a small mitogenic polypeptide present throughout a large number of tissues and body fluids in many mammalian species. EGF is a member of a growth factor family characterized by 6 conserved cysteine motifs that form three disulfide bonds. Human EGF is an identical molecule to ß-urogastrone, a molecule isolated on the basis of its ability to inhibit gastric acid secretion. EGF is structurally homologous to human transforming growth factor-a, and both exert their actions through EGF receptors.

Caution

This product should be stored at 2-8°C. After reconstitution, it can be stored at 2-8°C for two weeks, or frozen in aliquots at -20°C for longer periods.

Preparation Note

This product was lyophilized from a solution in 1 mL of 5 mM ammonium acetate at pH 6.5. It should be reconstituted by adding the contents of the vial to a solution containing 0.1-1.0% BSA or 1-10% serum in buffered saline or tissue culture medium.

Analysis Note

The biological activity of this product is measured by an ability to stimulate the EGF-responsive mouse BALB/MK cells. The EC50 is defined as the effective concentration of growth factor that elicits a 50% increase in cell growth in a cell based bioassay.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of amino acids on maturation, fertilization and embryo development of pig follicular oocytes in two IVM media
Hong JY, et al.
Theriogenology, 62(8), 1473-1482 (2004)
Expression and activation of mitogen-activated protein kinases in matured porcine oocytes under thermal stress
Yen SY, et al.
Journal of Reproduction and Development, 60(5), 388-394 (2014)
An efficient system for selection and culture of Schwann cells from adult rat peripheral nerves
Niapour N, et al.
Cytotechnology, 68(4), 629-636 (2016)
Yue Qian et al.
Developmental biology, 394(2), 217-227 (2014-08-30)
The development of stem cell daughters into the differentiated state normally requires a cascade of proliferation and differentiation steps that are typically regulated by external signals. The germline cells of most animals, in specific, are associated with somatic support cells
Xiaoni Li et al.
The Journal of biological chemistry, 288(25), 18121-18133 (2013-05-08)
Emerging evidence indicates that the miR-23a/24-2/27a cluster may possess a causal role in mammary tumorigenesis and function as a novel class of oncogenes. However, the regulatory mechanism of the miR-23a/24-2/27a cluster in mammary carcinoma cell invasion and migration is still

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service