SRP3089
IL-22 human
recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Synonym(s):
IL-TIF
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
biological source
human
recombinant
expressed in E. coli
Assay
≥98% (HPLC)
≥98% (SDS-PAGE)
form
lyophilized
mol wt
32.8 kDa
packaging
pkg of 10 μg
technique(s)
cell culture | mammalian: suitable
impurities
<0.1 EU/μg endotoxin, tested
color
white to off-white
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... IL22(50616)
General description
IL22 (interleukin 22) is a member of the IL10 related cytokine family, and this gene is localized to the human chromosome 12q15. This protein is predominantly produced by activated CD4+ T cells, and its target cells include mesangial, neuronal and hepatoma cell types. Recombinant human IL-22 is a 33.6 kDa non-disulfide-linked homodimeric protein containing of two 147 amino acid polypeptide chains.
Biochem/physiol Actions
IL22 (interleukin 22) is responsible for eliciting acute-phase responses including, anemia, weight loss, increase in platelet numbers, elevated serum amyloid A and fibrinogen levels and reduced levels serum albumin levels. During inflammation, this protein leads to an increase in pancreatitis-associated protein 1 (PAP1) and osteopontin in pancreatic acinar cells, which might either have protective or trophic effects. This protein is involved in the crosstalk between keratinocytes and inflammatory cells. It is linked with tumorigenesis and progression of tumors. 429 C/T polymorphism in this gene is linked with susceptibility to bladder cancer. In patients with recurrent pregnancy loss, the expression of IL22 is reduced in the uterine decidua, which might lead to disruption of decidual homeostasis, and hence, early loss of pregnancy.
Sequence
MAPISSHCRL DKSNFQQPYI TNRTFMLAKE ASLADNNTDV RLIGEKLFHG VSMSERCYLM KQVLNFTLEE VLFPQSDRFQ PYMQEVVPFL ARLSNRLSTC HIEGDDLHIQ RNVQKLKDTV KKLGESGEIK AIGELDLLFM SLRNACI
Physical form
Lyophilized from 10 mM NaCitrate, pH 3.0
Reconstitution
Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.5-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Regulation of T cells and cytokines by the interleukin-10 (IL-10)-family cytokines IL-19, IL-20, IL-22, IL-24 and IL-26.
Oral HB, et al.
European Journal of Immunology, 36(2), 380-388 (2006)
Expression of interleukin-22 in decidua of patients with early pregnancy and unexplained recurrent pregnancy loss.
O'Hern Perfetto C, et al.
Journal of Assisted Reproduction and Genetics, 32(6), 977-984 (2015)
Tao Zhao et al.
Clinics (Sao Paulo, Brazil), 70(10), 686-690 (2015-11-26)
The cytokine interleukin-22 (IL-22), which is produced by T cells and natural killer cells, is associated with tumorigenesis and tumor progression in cancers. However, the role of IL-22 in bladder cancer has not been investigated. A prospective hospital-based case-control study
Sergei V Kotenko
Cytokine & growth factor reviews, 13(3), 223-240 (2002-12-19)
Five novel cytokines (IL-19, IL-20, IL-22 (IL-TIF), IL-24 (human MDA-7, mouse FISP, rat C49A/Mob-5), and IL-26 (AK155)) demonstrating limited primary sequence identity and probable structural homology to IL-10 have been identified. These cellular cytokines, as well as several cytokines encoded
P Conti et al.
Immunology letters, 88(3), 171-174 (2003-08-28)
It has been reported that the CD4+ T cell is a very important source of interleukin 10 (IL-10), while CD8+ cells produce low amounts. IL-10 exerts several immune stimulating, as well as inhibitory effects. There are at least five novel
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service