Skip to Content
Merck
All Photos(2)

Key Documents

I1395

Sigma-Aldrich

IL-6 human

≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

Interleukin-6 human, hIL-6, IL-6

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Interleukin-6 human, IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

0.2-2.0 ng/mL ED50/EC50

quality

endotoxin tested

mol wt

26 kDa

packaging

pkg of 10 and 50 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

≤1.000 EU/μg

color

white

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL6(3569)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Interleukin-6 (IL-6) is a multifunctional protein originally discovered in the media of cells stimulated with double stranded RNA. IL-6 appears to be directly involved in the responses that occur after infection and injury and may prove to be as important as IL-1 and TNF-α in regulating the acute phase response. IL-6 is reported to be produced by fibroblasts, activated T cells, activated monocytes or macrophages, and endothelial cells. It acts upon a variety of cells, including fibroblasts, myeloid progenitor cells, T cells, B cells and hepatocytes. IL-6 induces multiple effects, as indicated by its numerous synonyms: plasmacytoma growth factor (PCT-GF), interferon-β-2 (IFN-β2), monocyte derived human B cell growth factor, B cell stimulating factor (BSF-2), hepatocyte stimulating factor (HSF), Interleukin Hybridoma/Plasmacytoma-1 (IL-HP1). In addition, IL-6 appears to interact with IL-2 in the proliferation of T lymphocytes. IL-6 also potentiates the proliferative effect of IL-3 on multipotential hematopoietic progenitors.

Physical form

Lyophilized from a 0.2 μm filtered solution of phosphate buffer saline (PBS), pH 7.4, containing 500 μg bovine serum albumin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Ferraiuolo et al.
Cancer letters, 433, 18-32 (2018-06-23)
Osteosarcoma (OS) is the most aggressive type of primary solid tumor that develops in bone. Whilst conventional chemotherapy can improve survival rates, the outcome for patients with metastatic or recurrent OS remains poor, so novel treatment agents and strategies are
Hemn Mohammadpour et al.
Cell reports, 37(4), 109883-109883 (2021-10-28)
Myeloid-derived suppressor cells (MDSCs) impede antitumor immunity; however, the precise mechanisms that regulate their suppressive function remain unresolved. Identifying these mechanisms could lead to therapeutic interventions to boost cancer immunotherapy efficacy. Here, we reveal that β2 adrenergic receptor (β2-AR) expression
Bin Zhang et al.
Stem cells and development, 23(11), 1233-1244 (2013-12-26)
Mesenchymal stem cells (MSCs) have been shown to secrete exosomes that are cardioprotective. Here, we demonstrated that MSC exosome, a secreted membrane vesicle, is immunologically active. MSC exosomes induced polymyxin-resistant, MYD88-dependent secreted embryonic alkaline phosphatase (SEAP) expression in a THP1-Xblue
Adriana Catalli et al.
PloS one, 9(5), e96891-e96891 (2014-05-14)
Despite the fact that glucocorticoids and long acting beta agonists are effective treatments for asthma, their effects on human mast cells (MC) appear to be modest. Although MC are one of the major effector cells in the underlying inflammatory reactions
Lei Wang et al.
Nature communications, 11(1), 5455-5455 (2020-10-30)
The expansion of the white adipose tissue (WAT) in obesity goes along with increased mechanical, metabolic and inflammatory stress. How adipocytes resist this stress is still poorly understood. Both in human and mouse adipocytes, the transcriptional co-activators YAP/TAZ and YAP/TAZ

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service