Skip to Content
Merck
All Photos(1)

Documents

SRP3058

Sigma-Aldrich

IFN-γ human

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

Immune Interferon, MAF, T cell interferon, type II interferon

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

5.0-10.0 ng/mL ED50

mol wt

16.7 kDa

packaging

pkg of 100 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IFNG(3458)

General description

IFN-γ (interferon γ) is an acid-labile glycosylated interferon produced by CD4 and CD8 T lymphocytes as well as activated NK (natural killer) cells. IFN-γ receptors are present in most immune cells, which respond to IFN-γ signaling by increasing the surface expression of class I MHC (major histocompatibility complex) proteins. This promotes the presentation of antigen to T-helper (CD4+) cells. The gene is mapped to human chromosome 12q15. Human IFN-γ is species-specific and is biologically active only in human and primate cells. Recombinant human IFN-γ is a 16.7kDa protein containing 143 amino acid residues.

Application

IFN-γ (interferon γ)-human has been used for the generation of inflammatory M1 cells from human monocytic cell line, THP1.

Biochem/physiol Actions

IFN-γ (interferon γ) signaling in antigen-presenting cells and antigen-recognizing B and T lymphocytes regulate the antigen-specific phases of the immune response. It can suppress the generation of anti-inflammatory cytokines and enhance the secretion of proinflammatory cytokines. Additionally, IFN-γ stimulates a number of lymphoid cell functions including the anti-microbial and anti-tumor responses of macrophages, NK (natural killer) cells and neutrophils. It is responsible for tumor rejection and is capable of killing tumor cells by autophagy or apoptosis. IFN-γ is also involved in Hepatitis E virus disease pathogenesis.

Sequence

MQDPYVKEAE NLKKYFNAGH SDVADNGTLF LGILKNWKEE SDRKIMQSQI VSFYFKLFKN FKDDQSIQKS VETIKEDMNV KFFNSNKKKR DDFEKLTNYS VTDLNVQRKA IHELIQVMAE LSPAAKTGKR KRSQMLFQGR RASQ

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.4.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a stabilizer (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Refined mapping of 1q32 amplicons in malignant gliomas confirms MDM4 as the main amplification target.
Riemenschneider MJ
International Journal of Cancer. Journal International Du Cancer, 104, 752-757 (2003)
Dynamic regulation of IFN-gamma signaling in antigen-specific CD8+ T cells responding to infection.
Haring JS
Journal of Immunology, 174, 6791-6802 (2005)
Distinct effects of T-bet in TH1 lineage commitment and IFN-gamma production in CD4 and CD8 T cells.
Szabo SJ
Science, 295, 338-342 (2002)
Interferons and their actions.
S Pestka et al.
Annual review of biochemistry, 56, 727-777 (1987-01-01)
Synergistic engagement of an ineffective endogenous anti-tumor immune response and induction of IFN-gamma and Fas-ligand-dependent tumor eradication by combined administration of IL-18 and IL-2.
Wigginton JM
Journal of Immunology, 169, 4467-4474 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service