Skip to Content
Merck
All Photos(2)

Key Documents

SML3957

Sigma-Aldrich

OSS_128167

≥98% (HPLC)

Synonym(s):

5-[[3-[(2-Furanylcarbonyl)amino]benzoyl]amino]-2-hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14N2O6
CAS Number:
Molecular Weight:
366.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

eCll penetrant, potent and selective inhibitor of SIRT6.



OSS_128167 is a cell penetrant, potent and selective inhibitor of SIRT6 (Sirtuin 6) that exhibits anti-cancer, anti-inflammatory, and anti-viral properties. OSS_128167 inhibits HBV transcription and replication through upregulating peroxisome proliferator-activated receptors α (PPARα) expression.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SIRT6 Inhibitor, OSS_128167 Restricts Hepatitis B Virus Transcription and Replication Through Targeting Transcription Factor Peroxisome Proliferator-Activated Receptors ?
Frontiers in Pharmacology, 10, 1270-1270 (2019)
Diosgenin protects against podocyte injury in early phase of diabetic nephropathy through regulating SIRT6
Phytomedicine, 104, 154276-154276 (2022)
Discovery of novel and selective SIRT6 inhibitors
Journal of Medicinal Chemistry, 57(11), 4796-4804 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service