Skip to Content
Merck
All Photos(1)

Key Documents

SML3242

Sigma-Aldrich

MR6-31-2

≥98% (HPLC)

Synonym(s):

2-(4-Chlorobenzyl)benzo[d][1,2]selenazol-3(2H)-one, 2-[(4-Chlorophenyl)methyl]-1,2-benzisoselenazol-3(2H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10ClNOSe
CAS Number:
Molecular Weight:
322.65
UNSPSC Code:
12352107
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

storage temp.

2-8°C

SMILES string

O=C1N(CC2=CC=C(Cl)C=C2)[Se]C3=CC=CC=C31

InChI

1S/C14H10ClNOSe/c15-11-7-5-10(6-8-11)9-16-14(17)12-3-1-2-4-13(12)18-16/h1-8H,9H2

InChI key

XZWKXIFLMYRVTK-UHFFFAOYSA-N

Biochem/physiol Actions

CNS penetrant potent inhibitor of SARS-CoV-2 Mpro that bind at the Mpro catalytic site
MR6-31-2, a CNS penetrant ebselen analog, is a potent inhibitor of SARS-CoV-2 Mpro that bind at the Mpro catalytic site. MR6-31-2 donates a selenium atom, forming a covalent bond and blocking the histidine-Cys catalytic dyad. It exhibits good neuroprotective effects and low cytotoxicity in cell-based and mouse models of motor neuron disease. MR6-31-2 covalently binds to A4V superoxide dismutase-1 (SOD1) and promotes its thermal stability.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kangsa Amporndanai et al.
Nature communications, 12(1), 3061-3061 (2021-05-26)
The SARS-CoV-2 pandemic has triggered global efforts to develop therapeutics. The main protease of SARS-CoV-2 (Mpro), critical for viral replication, is a key target for therapeutic development. An organoselenium drug called ebselen has been demonstrated to have potent Mpro inhibition and
Kangsa Amporndanai et al.
Nature communications, 12(1), 3061-3061 (2021-05-26)
The SARS-CoV-2 pandemic has triggered global efforts to develop therapeutics. The main protease of SARS-CoV-2 (Mpro), critical for viral replication, is a key target for therapeutic development. An organoselenium drug called ebselen has been demonstrated to have potent Mpro inhibition and
Varunya Chantadul et al.
Communications biology, 3(1), 97-97 (2020-03-07)
Mutations to the gene encoding superoxide dismutase-1 (SOD1) were the first genetic elements discovered that cause motor neuron disease (MND). These mutations result in compromised SOD1 dimer stability, with one of the severest and most common mutations Ala4Val (A4V) displaying
Varunya Chantadul et al.
Communications biology, 3(1), 97-97 (2020-03-07)
Mutations to the gene encoding superoxide dismutase-1 (SOD1) were the first genetic elements discovered that cause motor neuron disease (MND). These mutations result in compromised SOD1 dimer stability, with one of the severest and most common mutations Ala4Val (A4V) displaying

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service