Skip to Content
Merck
All Photos(1)

Key Documents

P1499

Sigma-Aldrich

Pterostilbene

≥97% (HPLC), solid

Synonym(s):

Pterocarpus marsupium, 3,5-Dimethoxy-4′-hydroxystilbene, 4-[(1E)-2-(3,5-Dimethoxyphenyl)ethenyl]phenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16O3
CAS Number:
Molecular Weight:
256.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

form

solid

storage condition

protect from light

solubility

DMSO: >20 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

COc1cc(OC)cc(\C=C\c2ccc(O)cc2)c1

InChI

1S/C16H16O3/c1-18-15-9-13(10-16(11-15)19-2)4-3-12-5-7-14(17)8-6-12/h3-11,17H,1-2H3/b4-3+

InChI key

VLEUZFDZJKSGMX-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pterostilbene (trans-3,5-dimethoxy-4-hydroxystilbene), an antioxidant is an analogue of resveratrol. It is present in blueberries, tree wood and some types of grapes.

Application

Pterostilbene has been used:
  • to investigate its anti-oxidative stress activities and the involvement of the nuclear factor (erythroid-derived 2)-like 2 (Nrf2)-antioxidant response element (ARE) signaling pathway
  • to determine its effects on transcriptional activation of estrogen receptor-α (ERα) in hormone resistant breast cancer cells)
  • to study its effects on the cytotoxicity of 2-chloroethyl ethyl sulphide (CEES) in human epidermoid carcinoma cells (A-431) through MTT (3-[4,5-dimethylthiazol-2-yl]-2,5 diphenyl tetrazolium bromide)viability assay

Biochem/physiol Actions

Antioxidant, antiproliferative, apoptosis inducer, antihyperglycemic, antidiabetic.
Pterostilbene exhibits antineoplastic properties in some of the malignancies, hence it is considered as an active anticancer agent. It has the ability to stimulate apoptosis and block the viability of cell in estrogen-receptor positive and negative breast cancer cell lines.

Caution

Air-sensitive

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ebselen analogues reduce 2-chloroethyl ethyl sulphide toxicity in A-431 cells
Pino M A, et al.
Arhiv za Higijenu Rada i Toksikologiju, 64(1), 77-86 (2013)
Victor Chia-Hsiang Lin et al.
Journal of agricultural and food chemistry, 60(25), 6399-6407 (2012-06-08)
Prostate cancer is one of the leading causes of cancer death in men in Western countries. Epidemiological studies have linked the consumption of fruits and vegetables to a reduced risk of prostate cancer, and small fruits are particularly rich sources
Wasamon Nutakul et al.
Journal of agricultural and food chemistry, 59(20), 10964-10970 (2011-09-23)
The effects of resveratrol and pterostilbene (two structurally related stilbene compounds) on three human colon cancer cells were systematically compared. Cell viability tests indicated that IC(50) values of pterostilbene were 2-5-fold lower than those of resveratrol in all three cancer
Hee Jeong Shin et al.
Molecules (Basel, Switzerland), 25(1) (2020-01-16)
Increasing studies have reported that cancer stem cells (CSCs) play critical roles in therapeutic resistance, recurrence, and metastasis of tumors, including cervical cancer. Pterostilbene, a dimethylated derivative of resveratrol, is a plant polyphenol compound with potential chemopreventive activity. However, the
Shih-Chun Yang et al.
Frontiers in microbiology, 8, 1103-1103 (2017-07-01)
Pterostilbene is a methoxylated derivative of resveratrol originated from natural sources. We investigated the antibacterial activity of pterostilbene against drug-resistant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service