Skip to Content
Merck

Skip To

MSCAL2

ProteoMass Peptide MALDI-MS Calibration Kit

Peptide MALDI MS Calibration Standard

Synonym(s):

Peptide Mass Spectrometry Calibrators

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

ProteoMass

Quality Segment

quality

Peptide MALDI MS Calibration Standard

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

storage temp.

15-25°C

Application

This kit features a convenient selection of pre-qualified standard peptides, matrices, and solvents for calibrating and testing matrix assisted laser desorption ionization (MALDI) mass spectrometers. Configured specifically for peptide applications (in the mass range of 700 to 3500 Da), MSCAL2 is ideal for analysis of tryptic fragments or post-source decay studies. Applications also include calibration, tuning, and sensitivity testing of MALDI instruments from all manufacturers.
ProteoMass Peptide MALDI-MS Calibration Kit has been used as a mass calibration standard for MALDI-TOF MS (matrix assisted laser desorption ionization-time of flight mass spectrometry) analysis.

Legal Information

ProteoMass is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MSCAL1MSCAL3C8857
description

Peptide MALDI MS Calibration Standard

description

Peptide and Protein MALDI MS Calibration Standard

description

Protein MALDI MS Calibration Standard

description

vial of 10 nmol, (M+H+) 12,361.96 Da by calculation

product line

ProteoMass

product line

ProteoMass

product line

ProteoMass

product line

ProteoMass

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

quality

Peptide MALDI MS Calibration Standard

quality

Peptide and Protein MALDI MS Calibration Standard

quality

Protein MALDI MS Calibration Standard

quality

-

storage temp.

15-25°C

storage temp.

-

storage temp.

15-25°C

storage temp.

-

MALDI calibration kit for sensitivity testing of mass spectrometers

2-isopropyl-5-methylbenzenethiol 98% purity, 2-isopropyl-5-methylbenzenethiol Sigma-Aldrich, 2-isopropyl-5-methylbenzenethiol analytical standards, 2-isopropyl-5-methylbenzenethiol applications in research, 2-isopropyl-5-methylbenzenethiol chemical structure, 2-isopropyl-5-methylbenzenethiol for laboratories, 2-isopropyl-5-methylbenzenethiol for organic synthesis, 2-isopropyl-5-methylbenzenethiol in chromatography, 2-isopropyl-5-methylbenzenethiol lab-grade, 2-isopropyl-5-methylbenzenethiol reagent grade, 2-isopropyl-5-methylbenzenethiol safety data sheet, 2-isopropyl-5-methylbenzenethiol synthesis methods, CAS 102-97-6 2-isopropyl-5-methylbenzenethiol

-

-


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ProteoMass Bradykinin Fragment 1-7 MALDI-MS Standard, vial of 10 nmol, monoisotopic mol wt 756.3997 Da 2 x 10
    SDS

  • ProteoMass Angiotensin II MALDI-MS Standard, vial of 10 nmol 2 x 10
    SDS

  • ProteoMass P14R MALDI-MS Standard, vial of 10 nmol 2 x 10
    SDS

  • ProteoMass ACTH Fragment 18-39 MALDI-MS Standard, vial of 10 nmol, monoisotopic mol wt 2,464.1989 Da 2 x 10
    SDS

  • ProteoMass Insulin chain B oxidized MALDI-MS Standard, vial of 10 nmol 2 x 10
    SDS

  • Acetonitrile 30 mL
    SDS

  • α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS 8 x 10
    SDS

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

3 - Flammable liquids

flash_point_f

35.6 °F

flash_point_c

2 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. ProteoMass™ Peptide MALDI-MS Calibration Kitに含まれるペプチドの名前とそららの分子量を教えてください。

    1 answer
    1. The components of this kit are listed below. Note that these are also published in the product SDS, section 16.
      A8346 – ACTH Fragment 18 – 39
      A8846 – Angiotensin II
      B4181 - Bradykinin Fragment 1 – 7
      C8982 – ?-Cyano-4-hydroxycinnamic acid
      I6154 – Insulin chain B, oxidized
      P2613 – P14R, (Proline) 14 Arginine
      A8596 – Acetonitrile
      T3693 - Trifluoroacetic acid solution, 1%
      T3443 – Trifluoroacetic acid solution, 0.1%

      Helpful?

Reviews

No rating value

Active Filters