Skip to Content
Merck
All Photos(1)

Key Documents

SML2955

Sigma-Aldrich

Mirodenafil dihydrochloride

≥98% (HPLC)

Synonym(s):

4-[[3-(5-Ethyl-4,5-dihydro-4-oxo-7-propyl-1H-pyrrolo[3,2-d]pyrimidin-2-yl)-4-propoxyphenyl]sulfonyl]-1-piperazineethanol, dihydrochloride, 5-Ethyl-2-[5-[[4-(2-hydroxyethyl)piperazin-1-yl]sulfonyl]-2-propoxyphenyl]-7-propyl-3,5-dihydro-4H-pyrrolo[3,2-d]pyrimidin-4-one, dihydrochloride, 5-Ethyl-3,5-dihydro-2-[5-[[4-(2-hydroxyethyl)-1-piperazinyl]sulfonyl]-2-propoxyphenyl]-7-propyl-4H-pyrrolo[3,2-d]pyrimidin-4-one, dihydrochloride, SK 3530 dihydrochloride, SK-3530 dihydrochloride, SK3530 dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H37N5O5S·2HCl
CAS Number:
Molecular Weight:
604.59
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[S](=O)(=O)(N4CCN(CC4)CCO)c1cc(c(cc1)OCCC)c2[nH][c](c3[n](cc(c3n2)CCC)CC)=O.Cl.Cl

InChI

1S/C26H37N5O5S.2ClH/c1-4-7-19-18-30(6-3)24-23(19)27-25(28-26(24)33)21-17-20(8-9-22(21)36-16-5-2)37(34,35)31-12-10-29(11-13-31)14-15-32;;/h8-9,17-18,32H,4-7,10-16H2,1-3H3,(H,27,28,33);2*1H

InChI key

CKPHITUXXABKDL-UHFFFAOYSA-N

Biochem/physiol Actions

Mirodenafil (SK3530) is an orally active, highly potent and selective phosphodiesterase-5 (PDE5) inhibitor (IC50 = 338 pM). with therapeutic efficacy in various ED models in vivo. Comparing to sildenafil, mirodenafil shows ~10-times higher PDE5 selectivity in vitro and remains longer in the plasma and corpus cavernosum following oral dosing at 40 mg/kg in rats in vivo.
Orally active, highly potent and selective phosphodiesterase-5 (PDE5) inhibitor with ED therapeutic efficacy in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Young H Choi et al.
Biopharmaceutics & drug disposition, 30(6), 305-317 (2009-07-30)
The pharmacokinetics of mirodenafil and its two metabolites, SK3541 and SK3544, after intravenous (5, 10, 20 and 50 mg/kg) and oral (10, 20 and 50 mg/kg) administration of mirodenafil, and the first-pass effect of mirodenafil after intravenous, oral, intraportal, intragastric
Ji-Youn Jung et al.
The Journal of veterinary medical science, 70(11), 1199-1204 (2008-12-06)
Mirodenafil (SK3530) is a new potent and selective inhibitor of cGMP-specific phosphodiesterase type 5 (PDE5). Recent clinical trials have demonstrated that mirodenafil is an effective treatment for erectile dysfunction. Its mechanism of action is enhancement of nitric oxide (NO) induced
Ji Sung Shim et al.
Urology, 91, 244-244 (2016-02-28)
To evaluate the distribution of a daily phosphodiesterase type 5 inhibitor dose (mirodenafil) in rat plasma and bladder and prostate tissue in a model of atherosclerosis-induced chronic pelvic ischemia. Thirty-two 18-week-old male Sprague Dawley rats were divided into two groups.
Jae Heon Kim et al.
The world journal of men's health, 38(3), 345-352 (2019-08-07)
To validate a novel arteriogenic erectile dysfunction (ED) model with atherosclerosis (AS) based on molecular and histologic evidence induced by chronic pelvic ischemia (CPI) and determine effect of phosphodiesterase-5 inhibitor treatment. Twenty 16-week-old male Sprague-Dawley rats were divided into three
Hoon Choi et al.
International neurourology journal, 19(1), 19-26 (2015-04-04)
To investigate the protective effect of mirodenafil on bladder function in a rat model of chronic bladder ischemia (CBI). Twenty-four Sprague-Dawley rats were randomized to three groups: untreated, sham-operated rats (control group); untreated, CBI model rats (CBI group); and CBI

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service