Skip to Content
Merck
All Photos(1)

Key Documents

SML2279

Sigma-Aldrich

VU0155069

≥98% (HPLC)

Synonym(s):

(S)-N-(1-(4-(5-Chloro-2-oxo-2,3-dihydro-1H-benzo[d]imidazol-1-yl)piperidin-1-yl)propan-2-yl)-2-naphthamide, CAY10593, N-[(1S)-2-[4-(5-Chloro-2,3-dihydro-2-oxo-1H-benzimidazol-1-yl)-1-piperidinyl]-1-methylethyl]-2-naphthalenecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H27ClN4O2
CAS Number:
Molecular Weight:
462.97
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

storage temp.

2-8°C

SMILES string

O=C(C1=CC=C2C=CC=CC2=C1)N[C@@H](C)CN3CCC(N4C5=CC=C(Cl)C=C5NC4=O)CC3

InChI

1S/C26H27ClN4O2.ClH/c1-17(28-25(32)20-7-6-18-4-2-3-5-19(18)14-20)16-30-12-10-22(11-13-30)31-24-9-8-21(27)15-23(24)29-26(31)33;/h2-9,14-15,17,22H,10-13,16H2,1H3,(H,28,32)(H,29,33);1H/t17-;/m0./s1

InChI key

RQULTQQAHGYYDG-LMOVPXPDSA-N

Biochem/physiol Actions

VU0155069 is a PLD1-selective catalytic site-targeting phospholipase D inhibitor (PLD1/2 IC50 = 46 nM/933 nM by cell-free enzymatic assays; substrate = di-palmitoyl-PLC) that selectively suppresses the cellular PLD activity in PMA-stimulated non-small-cell lung cancer (NSCLC) Calu-1 cells with predominant PLD1 activity over GFP-PLD2-overexpressing HEK293 cells (IC50 = 11 nM and 1.8 μM, respectively). Note: VU0155069 is reported to impair P2X7-induced pore formation in human RPMI 8226 B cells lacking PLD1 expression.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Woo Kang et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 23(23), 7340-7350 (2017-09-25)
Purpose: Dysregulated expression of PLD1 has emerged as a hallmark feature of colorectal cancer, which remains a major cause of mortality worldwide. Aberrant activation of Wnt/β-catenin signaling is a critical event in the development of colorectal cancer. Here, we investigated
Li-Da Luo et al.
Scientific reports, 7(1), 6035-6035 (2017-07-22)
Synapses are the basic units of information transmission, processing and integration in the nervous system. Dysfunction of the synaptic development has been recognized as one of the main reasons for mental dementia and psychiatric diseases such as Alzheimer's disease and
Dong Woo Kang et al.
Cancer research, 77(1), 142-152 (2016-10-30)
The RB1/E2F1 signaling pathway is frequently deregulated in colorectal cancer and has been suggested to intersect with Wnt/β-catenin and PI3K/Akt pathways, but molecular evidence for this link is lacking. In this study, we demonstrate that phospholipase D1 (PLD1), a transcriptional
A Pupovac et al.
Purinergic signalling, 9(4), 609-619 (2013-06-25)
The P2X7 receptor is a trimeric ATP-gated cation channel important in health and disease. We have observed that the specific phospholipase D (PLD)1 antagonist, CAY10593 impairs P2X7-induced shedding of the 'low affinity' IgE receptor, CD23. The current study investigated the
Hae-In Song et al.
Scientific reports, 6, 36968-36968 (2016-11-23)
Phospholipase D1 (PLD1) plays a known role in several differentiation processes, but its role in adipogenic differentiation remains unknown. In the present study, we identified PLD1 as a negative regulator of adipogenic differentiation. We showed that PLD activity was downregulated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service