Skip to Content
Merck
All Photos(1)

Key Documents

SML2200

Sigma-Aldrich

Octyl-(R)-2HG

≥98% (HPLC), film, α-KG-dependent dioxygenases prolyl hydroxylases inhibitor

Synonym(s):

(2R)-2-Hydroxyglutarate octyl ester, (2R)-Octyl-α-hydroxyglutarate, 1-Octyl-D-2-hydroxyglutarate, 1-octyl ester, 2R-Hydroxy-pentanedioic acid, Octyl-2HG, Octyl-D-2HG, R-2HG octyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H24O5
CAS Number:
Molecular Weight:
260.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Octyl-(R)-2HG, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

film

storage condition

desiccated

color

colorless

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O[C@H](CCC(=O)O)C(=O)OCCCCCCCC

InChI key

UJZOKTKSGUOCCM-LLVKDONJSA-N

Application

Octyl-(R)-2 hydroxyglutarate (HG) has been used as a membrane permeamble oncometabolite in gliblostoma cells to test its effects on NANOG transcription factor expression.[1] It has also been used as a competitive inhibitor of the enzyme α-ketoglutarate (α-KG)-dependent deoxygenase.[2]

Biochem/physiol Actions

Membrane-permeant precursor form of the oncometabolite D-2-hydroxyglutarate that inhibits α-ketoglutarate/α-KG-dependent dioxygenases.
Octyl-(R)-2HG (Octyl-D-2HG) is a membrane-permeant precursor form of the oncometabolite D-2-hydroxyglutarate (D-2HG) produced by tumor cells due to mutations in the NADP+-dependent isocitrate dehydrogenase genes IDH1 and IDH2. D-2HG inhibits multiple α-ketoglutarate/α-KG-dependent dioxygenases by competing against α-KG binding. Cellular D-2HG delivery by Octyl-(R)-2HG treatment (1-50 mM) is shown to suppress demethylase activity (~148% H3K9me2 and ~310% H3K79me2 upregulation; 50 mM in U-87MG) as well as increase HIF-1α and decrease endostatin levels as a result of inhibiting α-KG-dependent dioxygenases prolyl hydroxylases (PHDs) and collagen prolyl-4-hydroxylase (C-P4H), respectively.

Preparation Note

The density is not reported for this compound. The molecular weight is 260.33. The solubility is tested at 2mg/mL in DMSO, however other sources suggest that it is soluble in DMSO up to 10mg/mL.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zachary J Reitman et al.
The Journal of biological chemistry, 289(34), 23318-23328 (2014-07-06)
Mutations in the cytosolic NADP(+)-dependent isocitrate dehydrogenase (IDH1) occur in several types of cancer, and altered cellular metabolism associated with IDH1 mutations presents unique therapeutic opportunities. By altering IDH1, these mutations target a critical step in reductive glutamine metabolism, the
alpha-Ketoglutarate-Activated NF-$\kappa$B Signaling Promotes Compensatory Glucose Uptake and Brain Tumor Development
Wang X, et al.
Molecular Cell, 76(1), 148-162 (2019)
IDH1R132H Causes Resistance to HDAC Inhibitors by Increasing NANOG in Glioblastoma Cells
Kim G H, et al.
International Journal of Molecular Sciences, 20(11), 2679-2679 (2019)
Parker L Sulkowski et al.
Science translational medicine, 9(375) (2017-02-06)
2-Hydroxyglutarate (2HG) exists as two enantiomers, (R)-2HG and (S)-2HG, and both are implicated in tumor progression via their inhibitory effects on α-ketoglutarate (αKG)-dependent dioxygenases. The former is an oncometabolite that is induced by the neomorphic activity conferred by isocitrate dehydrogenase
Makoto Hirata et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(9), 2829-2834 (2015-03-03)
Enchondromas are benign cartilage tumors and precursors to malignant chondrosarcomas. Somatic mutations in the isocitrate dehydrogenase genes (IDH1 and IDH2) are present in the majority of these tumor types. How these mutations cause enchondromas is unclear. Here, we identified the

Questions

  1. What is the density of this product? I can´t find it and I want to have a 100mM stock.

    1 answer
    1. The density is not reported for this compound. The molecular weight is 260.33. The solubility is tested at 2mg/mL in DMSO, however other sources suggest that it is soluble in DMSO up to 10mg/mL. This rate has not been validated.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service