Skip to Content
Merck
All Photos(3)

Documents

H8378

Sigma-Aldrich

D-α-Hydroxyglutaric acid disodium salt

≥95% (GC), cell analysisfor peptide synthesis

Synonym(s):

(R)-2-Hydroxypentanedioic acid disodium salt, Disodium (R)-2-hydroxyglutarate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H6O5Na2
CAS Number:
Molecular Weight:
192.08
Beilstein:
5318041
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

product name

D-α-Hydroxyglutaric acid disodium salt, ≥95% (GC)

Assay

≥95% (GC)

form

powder

color

white to off-white

application(s)

cell analysis
peptide synthesis

storage temp.

−20°C

SMILES string

[Na].O[C@H](CCC(O)=O)C(O)=O

InChI

1S/C5H8O5.2Na/c6-3(5(9)10)1-2-4(7)8;;/h3,6H,1-2H2,(H,7,8)(H,9,10);;/q;2*+1/p-2/t3-;;/m1../s1

InChI key

DZHFTEDSQFPDPP-HWYNEVGZSA-L

Looking for similar products? Visit Product Comparison Guide

Application

D-α-Hydroxyglutaric acid disodium salt can be used as a reagent in peptide synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Hans et al.
European journal of biochemistry, 265(1), 404-414 (1999-09-22)
Component D (HgdAB) of 2-hydroxyglutaryl-CoA dehydratase from Clostridium symbiosum was purified to homogeneity. It is able to use component A from Acidaminococcus fermentans (HgdC) to initiate catalysis together with ATP, Mg2+ and a strong reducing agent such as Ti(III)citrate. Component
Postoperative risk of IDH mutant glioma-associated seizures and their potential management with IDH mutant inhibitors.
Drumm, et al.
The Journal of Clinical Investigation (2023)
The tumor suppressor prostate apoptosis response-4 (Par-4) is regulated by mutant IDH1 and kills glioma stem cells.
Liu Y, Gilbert MR, Kyprianou N, et al.
Acta Neuropathologica, 128(5), 723-732 (2014)
Shan Jin et al.
Nature communications, 14(1), 6638-6638 (2023-10-21)
Mammalian lactate dehydrogenase D (LDHD) catalyzes the oxidation of D-lactate to pyruvate. LDHD mutations identified in patients with D-lactic acidosis lead to deficient LDHD activity. Here, we perform a systematic biochemical study of mouse LDHD (mLDHD) and determine the crystal
Huan Wang et al.
International journal of cancer, 148(7), 1743-1755 (2020-12-16)
Metabolism reprograming is a hallmark of cancer and plays an important role in tumor progression. The aberrant metabolism in renal cell carcinoma (RCC) leads to accumulation of the oncometabolite L-2-hydroxyglurate (L-2HG). L-2HG has been reported to inhibit the activity of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service